Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38339508-38340012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPRED1
Entrez Gene ID
161742
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
NFLS, PPP1R147, hSpred1, spred-1
UniGene
Hs.525781
Gene Chromosome Location
Chr.15: 38252087-38357249 on Build GRCh38
Transcripts
NM_152594.2 XM_005254202.3 XM_011521289.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6019177 c.512C>G p.S171* Chr.15: 38339825-38339825 on Build GRCh38
5426214 c.424-8C>A p.? Chr.15: 38339729-38339729 on Build GRCh38
401538 c.473A>C p.Q158P Chr.15: 38339786-38339786 on Build GRCh38
3361606 c.545A>G p.N182S Chr.15: 38339858-38339858 on Build GRCh38
1135645 c.582+1G>T p.? Chr.15: 38339896-38339896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766431294 Chr.15: 38339891-38339891 on Build GRCh38
rs747565595 Chr.15: 38339694-38339694 on Build GRCh38
rs764990934 Chr.15: 38339880-38339880 on Build GRCh38
rs750421037 Chr.15: 38339764-38339764 on Build GRCh38
rs200142785 Chr.15: 38339878-38339878 on Build GRCh38
rs200632168 Chr.15: 38339918-38339918 on Build GRCh38
rs7163339 Chr.15: 38339639-38339639 on Build GRCh38
rs771673657 Chr.15: 38339823-38339823 on Build GRCh38
rs1013932170 Chr.15: 38339641-38339641 on Build GRCh38
rs781686509 Chr.15: 38339913-38339913 on Build GRCh38
rs369792420 Chr.15: 38339923-38339923 on Build GRCh38
rs769206547 Chr.15: 38339789-38339789 on Build GRCh38
rs553832516 Chr.15: 38339843-38339843 on Build GRCh38
rs202092705 Chr.15: 38339877-38339877 on Build GRCh38
rs903528084 Chr.15: 38339532-38339532 on Build GRCh38
rs370735097 Chr.15: 38339876-38339876 on Build GRCh38
rs200904257 Chr.15: 38339912-38339912 on Build GRCh38
rs955407365 Chr.15: 38339564-38339564 on Build GRCh38
rs370397468 Chr.15: 38339919-38339919 on Build GRCh38
rs759113238 Chr.15: 38339721-38339721 on Build GRCh38
rs1000577968 Chr.15: 38339550-38339550 on Build GRCh38
rs143121426 Chr.15: 38339848-38339848 on Build GRCh38
rs757989919 Chr.15: 38339690-38339690 on Build GRCh38
rs994188615 Chr.15: 38339693-38339693 on Build GRCh38
rs7179118 Chr.15: 38339719-38339719 on Build GRCh38
rs763971890 Chr.15: 38339737-38339737 on Build GRCh38
rs563635712 Chr.15: 38339627-38339627 on Build GRCh38
rs376527959 Chr.15: 38339824-38339824 on Build GRCh38
rs924180599 Chr.15: 38339625-38339625 on Build GRCh38
rs761239963 Chr.15: 38339648-38339648 on Build GRCh38
rs753701682 Chr.15: 38339739-38339739 on Build GRCh38
rs894877232 Chr.15: 38339953-38339953 on Build GRCh38
rs1016888945 Chr.15: 38339683-38339683 on Build GRCh38
rs894949375 Chr.15: 38339617-38339617 on Build GRCh38
rs759158378 Chr.15: 38339820-38339820 on Build GRCh38
rs763365429 Chr.15: 38339837-38339839 on Build GRCh38
rs1057518328 Chr.15: 38339733-38339733 on Build GRCh38
rs201335683 Chr.15: 38339874-38339874 on Build GRCh38
rs946857479 Chr.15: 38339618-38339618 on Build GRCh38
rs1044011965 Chr.15: 38339621-38339621 on Build GRCh38
rs773763661 Chr.15: 38339704-38339704 on Build GRCh38
rs370969521 Chr.15: 38339632-38339632 on Build GRCh38
rs1013790807 Chr.15: 38339979-38339979 on Build GRCh38
rs772419924 Chr.15: 38339687-38339687 on Build GRCh38
rs7181472 Chr.15: 38339947-38339947 on Build GRCh38
rs905722843 Chr.15: 38339665-38339665 on Build GRCh38
rs202069514 Chr.15: 38339705-38339705 on Build GRCh38
rs758617124 Chr.15: 38339883-38339883 on Build GRCh38
rs758483715 Chr.15: 38339776-38339776 on Build GRCh38
rs147159028 Chr.15: 38339688-38339688 on Build GRCh38
rs934383493 Chr.15: 38339626-38339626 on Build GRCh38
rs376549743 Chr.15: 38339594-38339594 on Build GRCh38
rs1005788400 Chr.15: 38339669-38339669 on Build GRCh38
rs768979377 Chr.15: 38339701-38339701 on Build GRCh38
rs749638449 Chr.15: 38339951-38339951 on Build GRCh38
rs781477854 Chr.15: 38339790-38339790 on Build GRCh38
rs770630398 Chr.15: 38339708-38339708 on Build GRCh38
rs1044229463 Chr.15: 38339647-38339647 on Build GRCh38
rs762498323 Chr.15: 38339707-38339707 on Build GRCh38
rs757357920 Chr.15: 38339751-38339751 on Build GRCh38
rs747508368 Chr.15: 38339783-38339783 on Build GRCh38
rs774070689 Chr.15: 38339816-38339816 on Build GRCh38
rs756609651 Chr.15: 38339922-38339922 on Build GRCh38
rs924918197 Chr.15: 38339932-38339932 on Build GRCh38
rs544963512 Chr.15: 38339609-38339609 on Build GRCh38
rs748550020 Chr.15: 38339808-38339808 on Build GRCh38
rs532147114 Chr.15: 38339832-38339832 on Build GRCh38
rs373384814 Chr.15: 38339759-38339759 on Build GRCh38
rs752656697 Chr.15: 38339732-38339732 on Build GRCh38
rs757914655 Chr.15: 38339960-38339961 on Build GRCh38
rs7180446 Chr.15: 38339729-38339729 on Build GRCh38
rs770446191 Chr.15: 38339811-38339811 on Build GRCh38
rs374835168 Chr.15: 38339921-38339921 on Build GRCh38
rs372086721 Chr.15: 38339949-38339949 on Build GRCh38
rs1052682360 Chr.15: 38339635-38339635 on Build GRCh38
rs764135694 Chr.15: 38339873-38339873 on Build GRCh38
rs376944837 Chr.15: 38339778-38339778 on Build GRCh38
rs372751409 Chr.15: 38339924-38339924 on Build GRCh38
rs375191510 Chr.15: 38339693-38339694 on Build GRCh38
rs751649950 Chr.15: 38339910-38339910 on Build GRCh38
rs374643618 Chr.15: 38339546-38339547 on Build GRCh38
rs921335586 Chr.15: 38339866-38339866 on Build GRCh38
rs143861527 Chr.15: 38339691-38339691 on Build GRCh38
rs752352987 Chr.15: 38339664-38339664 on Build GRCh38
rs962705113 Chr.15: 38339749-38339749 on Build GRCh38
rs760732248 Chr.15: 38339734-38339734 on Build GRCh38

View Full Product Details