Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18785236-18785753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMP
Entrez Gene ID
1311
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
UniGene
Hs.1584
Gene Chromosome Location
Chr.19: 18782773-18791305 on Build GRCh38
Transcripts
NM_000095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6150067 c.1717G>T p.G573C Chr.19: 18785498-18785498 on Build GRCh38
3100450 c.1690A>G p.M564V Chr.19: 18785525-18785525 on Build GRCh38
3712861 c.1634C>T p.A545V Chr.19: 18785707-18785707 on Build GRCh38
5693487 c.1664A>G p.N555S Chr.19: 18785677-18785677 on Build GRCh38
6250889 c.1611C>A p.V537V Chr.19: 18785730-18785730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749001252 Chr.19: 18785628-18785628 on Build GRCh38
rs750142844 Chr.19: 18785702-18785702 on Build GRCh38
rs952408771 Chr.19: 18785268-18785268 on Build GRCh38
rs757251084 Chr.19: 18785584-18785584 on Build GRCh38
rs1049783315 Chr.19: 18785449-18785449 on Build GRCh38
rs1048563479 Chr.19: 18785492-18785492 on Build GRCh38
rs758283232 Chr.19: 18785467-18785467 on Build GRCh38
rs200870935 Chr.19: 18785707-18785707 on Build GRCh38
rs534971433 Chr.19: 18785363-18785363 on Build GRCh38
rs202087711 Chr.19: 18785583-18785583 on Build GRCh38
rs750805331 Chr.19: 18785557-18785557 on Build GRCh38
rs773400537 Chr.19: 18785649-18785649 on Build GRCh38
rs762472383 Chr.19: 18785542-18785542 on Build GRCh38
rs546024072 Chr.19: 18785554-18785554 on Build GRCh38
rs868086369 Chr.19: 18785651-18785651 on Build GRCh38
rs911339664 Chr.19: 18785460-18785460 on Build GRCh38
rs749401915 Chr.19: 18785466-18785466 on Build GRCh38
rs780780400 Chr.19: 18785589-18785589 on Build GRCh38
rs751920575 Chr.19: 18785451-18785451 on Build GRCh38
rs751846896 Chr.19: 18785509-18785509 on Build GRCh38
rs199591164 Chr.19: 18785569-18785569 on Build GRCh38
rs983206008 Chr.19: 18785276-18785276 on Build GRCh38
rs765296978 Chr.19: 18785669-18785669 on Build GRCh38
rs1043943664 Chr.19: 18785696-18785696 on Build GRCh38
rs769488967 Chr.19: 18785517-18785517 on Build GRCh38
rs757668593 Chr.19: 18785455-18785455 on Build GRCh38
rs561997210 Chr.19: 18785597-18785597 on Build GRCh38
rs951397973 Chr.19: 18785627-18785627 on Build GRCh38
rs373548140 Chr.19: 18785596-18785596 on Build GRCh38
rs768082784 Chr.19: 18785643-18785643 on Build GRCh38
rs761134568 Chr.19: 18785650-18785650 on Build GRCh38
rs929885502 Chr.19: 18785321-18785322 on Build GRCh38
rs113138432 Chr.19: 18785317-18785317 on Build GRCh38
rs1027161629 Chr.19: 18785262-18785262 on Build GRCh38
rs779012253 Chr.19: 18785573-18785573 on Build GRCh38
rs145290786 Chr.19: 18785653-18785653 on Build GRCh38
rs942471982 Chr.19: 18785399-18785399 on Build GRCh38
rs369097923 Chr.19: 18785635-18785635 on Build GRCh38
rs1019950810 Chr.19: 18785602-18785602 on Build GRCh38
rs768047444 Chr.19: 18785458-18785458 on Build GRCh38
rs201160345 Chr.19: 18785471-18785471 on Build GRCh38
rs912211517 Chr.19: 18785369-18785369 on Build GRCh38
rs913131513 Chr.19: 18785410-18785410 on Build GRCh38
rs373498615 Chr.19: 18785691-18785691 on Build GRCh38
rs74432818 Chr.19: 18785660-18785660 on Build GRCh38
rs557134436 Chr.19: 18785465-18785465 on Build GRCh38
rs762347570 Chr.19: 18785637-18785637 on Build GRCh38
rs772420147 Chr.19: 18785723-18785723 on Build GRCh38
rs962493689 Chr.19: 18785263-18785263 on Build GRCh38
rs375299091 Chr.19: 18785624-18785624 on Build GRCh38
rs772859205 Chr.19: 18785397-18785397 on Build GRCh38
rs369668953 Chr.19: 18785561-18785561 on Build GRCh38
rs566112479 Chr.19: 18785709-18785709 on Build GRCh38
rs1041411983 Chr.19: 18785662-18785662 on Build GRCh38
rs939021336 Chr.19: 18785354-18785354 on Build GRCh38
rs1014333535 Chr.19: 18785571-18785571 on Build GRCh38
rs397515511 Chr.19: 18785676-18785676 on Build GRCh38
rs776545314 Chr.19: 18785510-18785510 on Build GRCh38
rs558677633 Chr.19: 18785275-18785275 on Build GRCh38
rs750600209 Chr.19: 18785459-18785459 on Build GRCh38
rs757080842 Chr.19: 18785685-18785685 on Build GRCh38
rs994431579 Chr.19: 18785483-18785483 on Build GRCh38
rs574375960 Chr.19: 18785425-18785425 on Build GRCh38
rs761133261 Chr.19: 18785727-18785727 on Build GRCh38
rs764701144 Chr.19: 18785467-18785468 on Build GRCh38
rs752778447 Chr.19: 18785672-18785672 on Build GRCh38
rs188276215 Chr.19: 18785417-18785417 on Build GRCh38
rs777719687 Chr.19: 18785468-18785468 on Build GRCh38
rs992533802 Chr.19: 18785279-18785279 on Build GRCh38
rs967666219 Chr.19: 18785605-18785605 on Build GRCh38
rs774801993 Chr.19: 18785519-18785519 on Build GRCh38
rs764652826 Chr.19: 18785538-18785538 on Build GRCh38
rs745684634 Chr.19: 18785592-18785592 on Build GRCh38
rs748950117 Chr.19: 18785648-18785648 on Build GRCh38
rs774980274 Chr.19: 18785541-18785541 on Build GRCh38
rs978668554 Chr.19: 18785615-18785615 on Build GRCh38
rs978526233 Chr.19: 18785424-18785424 on Build GRCh38
rs770690742 Chr.19: 18785486-18785486 on Build GRCh38
rs1012762778 Chr.19: 18785558-18785561 on Build GRCh38
rs779774621 Chr.19: 18785708-18785708 on Build GRCh38
rs925802600 Chr.19: 18785434-18785434 on Build GRCh38
rs999766204 Chr.19: 18785464-18785464 on Build GRCh38
rs779812541 Chr.19: 18785637-18785637 on Build GRCh38
rs113658931 Chr.19: 18785406-18785406 on Build GRCh38
rs1039434097 Chr.19: 18785403-18785403 on Build GRCh38
rs757719331 Chr.19: 18785596-18785596 on Build GRCh38
rs747684271 Chr.19: 18785722-18785722 on Build GRCh38
rs35701632 Chr.19: 18785277-18785277 on Build GRCh38
rs979859648 Chr.19: 18785271-18785271 on Build GRCh38
rs766649088 Chr.19: 18785565-18785565 on Build GRCh38
rs768270282 Chr.19: 18785630-18785630 on Build GRCh38
rs376678596 Chr.19: 18785524-18785524 on Build GRCh38
rs532716176 Chr.19: 18785443-18785443 on Build GRCh38
rs897362995 Chr.19: 18785475-18785475 on Build GRCh38
rs577011338 Chr.19: 18785513-18785513 on Build GRCh38
rs773740227 Chr.19: 18785724-18785724 on Build GRCh38

View Full Product Details