Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27367939-27368443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3682627 c.651C>G p.S217R Chr.2: 27368139-27368139 on Build GRCh38
3839280 c.544C>T p.H182Y Chr.2: 27368418-27368418 on Build GRCh38
5505724 c.653T>G p.L218R Chr.2: 27368077-27368077 on Build GRCh38
4924517 c.747G>T p.L249L Chr.2: 27368043-27368043 on Build GRCh38
6652720 c.669C>T p.I223I Chr.2: 27368121-27368121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990557179 Chr.2: 27367975-27367975 on Build GRCh38
rs752818834 Chr.2: 27368379-27368379 on Build GRCh38
rs1050304821 Chr.2: 27368208-27368208 on Build GRCh38
rs770574434 Chr.2: 27368082-27368082 on Build GRCh38
rs760641987 Chr.2: 27368102-27368102 on Build GRCh38
rs751780070 Chr.2: 27368152-27368152 on Build GRCh38
rs750664457 Chr.2: 27367977-27367977 on Build GRCh38
rs113994027 Chr.2: 27368047-27368047 on Build GRCh38
rs756090304 Chr.2: 27368392-27368392 on Build GRCh38
rs766717475 Chr.2: 27368136-27368136 on Build GRCh38
rs113994029 Chr.2: 27368105-27368105 on Build GRCh38
rs972532369 Chr.2: 27367968-27367968 on Build GRCh38
rs113309679 Chr.2: 27368320-27368320 on Build GRCh38
rs1008225417 Chr.2: 27368302-27368302 on Build GRCh38
rs761726266 Chr.2: 27368351-27368351 on Build GRCh38
rs757694836 Chr.2: 27368056-27368056 on Build GRCh38
rs199682969 Chr.2: 27368188-27368188 on Build GRCh38
rs753903529 Chr.2: 27368001-27368002 on Build GRCh38
rs748998115 Chr.2: 27368076-27368076 on Build GRCh38
rs775331791 Chr.2: 27368100-27368100 on Build GRCh38
rs781570273 Chr.2: 27368046-27368046 on Build GRCh38
rs768638665 Chr.2: 27368346-27368346 on Build GRCh38
rs751690179 Chr.2: 27367989-27367989 on Build GRCh38
rs1053260361 Chr.2: 27368240-27368240 on Build GRCh38
rs373696834 Chr.2: 27368352-27368352 on Build GRCh38
rs772991139 Chr.2: 27368390-27368390 on Build GRCh38
rs535744717 Chr.2: 27368332-27368332 on Build GRCh38
rs767601647 Chr.2: 27368167-27368167 on Build GRCh38
rs746252111 Chr.2: 27368288-27368288 on Build GRCh38
rs753667257 Chr.2: 27368130-27368130 on Build GRCh38
rs756631265 Chr.2: 27368181-27368181 on Build GRCh38
rs369686466 Chr.2: 27368324-27368324 on Build GRCh38
rs755137090 Chr.2: 27368156-27368156 on Build GRCh38
rs949043662 Chr.2: 27368021-27368021 on Build GRCh38
rs753194689 Chr.2: 27368179-27368179 on Build GRCh38
rs879271655 Chr.2: 27368415-27368415 on Build GRCh38
rs374915492 Chr.2: 27368322-27368322 on Build GRCh38
rs200307171 Chr.2: 27368138-27368138 on Build GRCh38
rs907630897 Chr.2: 27367974-27367974 on Build GRCh38
rs758683857 Chr.2: 27367980-27367980 on Build GRCh38
rs560111940 Chr.2: 27368091-27368091 on Build GRCh38
rs776656241 Chr.2: 27368350-27368350 on Build GRCh38
rs71441072 Chr.2: 27368242-27368242 on Build GRCh38
rs375458330 Chr.2: 27368396-27368396 on Build GRCh38
rs757657538 Chr.2: 27368416-27368416 on Build GRCh38
rs542713478 Chr.2: 27368312-27368314 on Build GRCh38
rs757260632 Chr.2: 27368183-27368184 on Build GRCh38
rs113994028 Chr.2: 27368104-27368104 on Build GRCh38
rs561112017 Chr.2: 27368277-27368277 on Build GRCh38
rs916241714 Chr.2: 27367979-27367979 on Build GRCh38
rs376346942 Chr.2: 27368073-27368073 on Build GRCh38
rs944048086 Chr.2: 27368235-27368235 on Build GRCh38
rs746981613 Chr.2: 27368333-27368333 on Build GRCh38
rs778566686 Chr.2: 27368323-27368323 on Build GRCh38
rs907448099 Chr.2: 27368057-27368057 on Build GRCh38
rs780352685 Chr.2: 27367988-27367988 on Build GRCh38
rs577067377 Chr.2: 27367969-27367969 on Build GRCh38
rs773935313 Chr.2: 27368089-27368089 on Build GRCh38
rs1012086936 Chr.2: 27368261-27368261 on Build GRCh38
rs778177199 Chr.2: 27368075-27368075 on Build GRCh38
rs756334195 Chr.2: 27368317-27368317 on Build GRCh38
rs1014356484 Chr.2: 27368341-27368341 on Build GRCh38
rs911189093 Chr.2: 27368227-27368227 on Build GRCh38
rs990980109 Chr.2: 27368361-27368361 on Build GRCh38
rs889786473 Chr.2: 27368280-27368281 on Build GRCh38
rs748646310 Chr.2: 27368048-27368048 on Build GRCh38
rs768826695 Chr.2: 27368342-27368342 on Build GRCh38
rs768451187 Chr.2: 27368175-27368175 on Build GRCh38
rs755047066 Chr.2: 27368030-27368030 on Build GRCh38
rs376400542 Chr.2: 27368330-27368330 on Build GRCh38
rs191734334 Chr.2: 27368355-27368355 on Build GRCh38
rs904731151 Chr.2: 27368319-27368319 on Build GRCh38
rs767553031 Chr.2: 27368375-27368375 on Build GRCh38
rs572469370 Chr.2: 27368219-27368219 on Build GRCh38

View Full Product Details