Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181238009-181238519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
UniGene
Hs.5662
Gene Chromosome Location
Chr.5: 181236928-181243906 on Build GRCh38
Transcripts
NM_006098.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1066941 c.774C>A p.I258I Chr.5: 181238102-181238102 on Build GRCh38
3013129 c.749C>T p.A250V Chr.5: 181238127-181238127 on Build GRCh38
5995991 c.715C>T p.L239L Chr.5: 181238161-181238161 on Build GRCh38
3615053 c.733C>T p.R245C Chr.5: 181238143-181238143 on Build GRCh38
7428474 c.753C>T p.A251A Chr.5: 181238123-181238123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776071682 Chr.5: 181238186-181238186 on Build GRCh38
rs768397876 Chr.5: 181238088-181238088 on Build GRCh38
rs572388060 Chr.5: 181238463-181238463 on Build GRCh38
rs762648527 Chr.5: 181238064-181238064 on Build GRCh38
rs750954171 Chr.5: 181238061-181238061 on Build GRCh38
rs187751892 Chr.5: 181238399-181238399 on Build GRCh38
rs774123881 Chr.5: 181238090-181238091 on Build GRCh38
rs2546422 Chr.5: 181238490-181238490 on Build GRCh38
rs1131424 Chr.5: 181238102-181238102 on Build GRCh38
rs570704731 Chr.5: 181238341-181238341 on Build GRCh38
rs746445861 Chr.5: 181238171-181238171 on Build GRCh38
rs1011725751 Chr.5: 181238263-181238263 on Build GRCh38
rs994098042 Chr.5: 181238316-181238316 on Build GRCh38
rs1040406651 Chr.5: 181238100-181238100 on Build GRCh38
rs533588663 Chr.5: 181238322-181238322 on Build GRCh38
rs529202308 Chr.5: 181238406-181238410 on Build GRCh38
rs548035538 Chr.5: 181238048-181238048 on Build GRCh38
rs150026038 Chr.5: 181238458-181238458 on Build GRCh38
rs767877625 Chr.5: 181238076-181238076 on Build GRCh38
rs2770997 Chr.5: 181238065-181238065 on Build GRCh38
rs756226455 Chr.5: 181238082-181238082 on Build GRCh38
rs11540198 Chr.5: 181238227-181238227 on Build GRCh38
rs745683332 Chr.5: 181238162-181238162 on Build GRCh38
rs113323374 Chr.5: 181238242-181238242 on Build GRCh38
rs749239716 Chr.5: 181238092-181238092 on Build GRCh38
rs115830226 Chr.5: 181238301-181238301 on Build GRCh38
rs976531032 Chr.5: 181238429-181238429 on Build GRCh38
rs201404764 Chr.5: 181238168-181238168 on Build GRCh38
rs201248084 Chr.5: 181238260-181238260 on Build GRCh38
rs11540197 Chr.5: 181238112-181238112 on Build GRCh38
rs767043301 Chr.5: 181238231-181238231 on Build GRCh38
rs773666887 Chr.5: 181238452-181238452 on Build GRCh38
rs57779325 Chr.5: 181238084-181238084 on Build GRCh38
rs763524329 Chr.5: 181238210-181238210 on Build GRCh38
rs889917567 Chr.5: 181238077-181238077 on Build GRCh38
rs540876120 Chr.5: 181238497-181238497 on Build GRCh38
rs752980640 Chr.5: 181238270-181238270 on Build GRCh38
rs896780641 Chr.5: 181238306-181238306 on Build GRCh38
rs182627488 Chr.5: 181238397-181238397 on Build GRCh38
rs755230976 Chr.5: 181238073-181238073 on Build GRCh38
rs964860891 Chr.5: 181238426-181238426 on Build GRCh38
rs1026878891 Chr.5: 181238317-181238317 on Build GRCh38
rs759772198 Chr.5: 181238373-181238373 on Build GRCh38
rs11540200 Chr.5: 181238123-181238123 on Build GRCh38
rs765595020 Chr.5: 181238055-181238055 on Build GRCh38
rs555899656 Chr.5: 181238401-181238401 on Build GRCh38
rs187034782 Chr.5: 181238180-181238180 on Build GRCh38
rs1042177764 Chr.5: 181238045-181238045 on Build GRCh38
rs780908382 Chr.5: 181238455-181238455 on Build GRCh38
rs148390201 Chr.5: 181238410-181238411 on Build GRCh38
rs370505076 Chr.5: 181238271-181238271 on Build GRCh38
rs2770998 Chr.5: 181238438-181238438 on Build GRCh38
rs922576949 Chr.5: 181238409-181238409 on Build GRCh38
rs776184981 Chr.5: 181238396-181238396 on Build GRCh38
rs554014729 Chr.5: 181238330-181238330 on Build GRCh38
rs771671631 Chr.5: 181238165-181238165 on Build GRCh38
rs193125580 Chr.5: 181238410-181238410 on Build GRCh38
rs189953976 Chr.5: 181238238-181238238 on Build GRCh38
rs752683404 Chr.5: 181238081-181238081 on Build GRCh38
rs761851164 Chr.5: 181238167-181238169 on Build GRCh38
rs767509519 Chr.5: 181238269-181238270 on Build GRCh38
rs146608631 Chr.5: 181238469-181238469 on Build GRCh38
rs185614235 Chr.5: 181238466-181238466 on Build GRCh38
rs568961277 Chr.5: 181238132-181238132 on Build GRCh38
rs760261932 Chr.5: 181238052-181238052 on Build GRCh38
rs1042292202 Chr.5: 181238393-181238393 on Build GRCh38
rs370355975 Chr.5: 181238063-181238063 on Build GRCh38
rs1044530147 Chr.5: 181238267-181238267 on Build GRCh38
rs544440619 Chr.5: 181238261-181238261 on Build GRCh38
rs778525909 Chr.5: 181238050-181238050 on Build GRCh38
rs959181410 Chr.5: 181238309-181238310 on Build GRCh38
rs893324925 Chr.5: 181238205-181238205 on Build GRCh38
rs11540181 Chr.5: 181238124-181238124 on Build GRCh38
rs145995963 Chr.5: 181238239-181238239 on Build GRCh38
rs943433838 Chr.5: 181238091-181238091 on Build GRCh38
rs746346568 Chr.5: 181238298-181238298 on Build GRCh38
rs767785052 Chr.5: 181238269-181238269 on Build GRCh38

View Full Product Details