Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37850258-37850748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1089437 c.91+1G>A p.? Chr.7: 37850436-37850436 on Build GRCh38
1450587 c.177C>T p.D59D Chr.7: 37850714-37850714 on Build GRCh38
5003652 c.129A>C p.R43S Chr.7: 37850666-37850666 on Build GRCh38
6489576 c.130G>A p.A44T Chr.7: 37850667-37850667 on Build GRCh38
1450586 c.9_10insA p.R6fs*11 Chr.7: 37850275-37850276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765556103 Chr.7: 37850693-37850694 on Build GRCh38
rs555707678 Chr.7: 37850336-37850336 on Build GRCh38
rs2722372 Chr.7: 37850665-37850665 on Build GRCh38
rs771877216 Chr.7: 37850448-37850448 on Build GRCh38
rs750902815 Chr.7: 37850378-37850378 on Build GRCh38
rs751539349 Chr.7: 37850721-37850721 on Build GRCh38
rs373353542 Chr.7: 37850291-37850291 on Build GRCh38
rs760905416 Chr.7: 37850682-37850682 on Build GRCh38
rs750630112 Chr.7: 37850417-37850417 on Build GRCh38
rs906219343 Chr.7: 37850516-37850516 on Build GRCh38
rs752092011 Chr.7: 37850401-37850401 on Build GRCh38
rs765880975 Chr.7: 37850369-37850369 on Build GRCh38
rs772021065 Chr.7: 37850357-37850357 on Build GRCh38
rs374519602 Chr.7: 37850470-37850470 on Build GRCh38
rs757623095 Chr.7: 37850300-37850300 on Build GRCh38
rs369447226 Chr.7: 37850593-37850593 on Build GRCh38
rs754089157 Chr.7: 37850699-37850699 on Build GRCh38
rs778106704 Chr.7: 37850296-37850296 on Build GRCh38
rs924858337 Chr.7: 37850391-37850391 on Build GRCh38
rs748116097 Chr.7: 37850445-37850445 on Build GRCh38
rs141405304 Chr.7: 37850450-37850450 on Build GRCh38
rs200758390 Chr.7: 37850632-37850632 on Build GRCh38
rs201220646 Chr.7: 37850388-37850388 on Build GRCh38
rs369098657 Chr.7: 37850297-37850297 on Build GRCh38
rs764751735 Chr.7: 37850475-37850475 on Build GRCh38
rs369542497 Chr.7: 37850709-37850709 on Build GRCh38
rs1050366765 Chr.7: 37850447-37850447 on Build GRCh38
rs758333100 Chr.7: 37850604-37850604 on Build GRCh38
rs777532091 Chr.7: 37850715-37850715 on Build GRCh38
rs375751041 Chr.7: 37850580-37850580 on Build GRCh38
rs373621005 Chr.7: 37850596-37850596 on Build GRCh38
rs202008099 Chr.7: 37850603-37850603 on Build GRCh38
rs757570040 Chr.7: 37850404-37850404 on Build GRCh38
rs753041716 Chr.7: 37850698-37850699 on Build GRCh38
rs774819055 Chr.7: 37850476-37850476 on Build GRCh38
rs760293308 Chr.7: 37850368-37850368 on Build GRCh38
rs772869964 Chr.7: 37850364-37850364 on Build GRCh38
rs770546236 Chr.7: 37850453-37850453 on Build GRCh38
rs766441690 Chr.7: 37850582-37850582 on Build GRCh38
rs138311368 Chr.7: 37850663-37850663 on Build GRCh38
rs763436203 Chr.7: 37850386-37850386 on Build GRCh38
rs868708838 Chr.7: 37850700-37850700 on Build GRCh38
rs756982461 Chr.7: 37850723-37850723 on Build GRCh38
rs559053210 Chr.7: 37850499-37850499 on Build GRCh38
rs753590172 Chr.7: 37850296-37850296 on Build GRCh38
rs372678302 Chr.7: 37850318-37850318 on Build GRCh38
rs776071598 Chr.7: 37850380-37850380 on Build GRCh38
rs1012630221 Chr.7: 37850507-37850507 on Build GRCh38
rs372718997 Chr.7: 37850486-37850486 on Build GRCh38
rs188238656 Chr.7: 37850311-37850311 on Build GRCh38
rs540938955 Chr.7: 37850636-37850636 on Build GRCh38
rs377033672 Chr.7: 37850320-37850320 on Build GRCh38
rs762350734 Chr.7: 37850477-37850477 on Build GRCh38
rs780172047 Chr.7: 37850426-37850426 on Build GRCh38
rs765828426 Chr.7: 37850290-37850290 on Build GRCh38
rs756360013 Chr.7: 37850418-37850418 on Build GRCh38
rs766786157 Chr.7: 37850685-37850685 on Build GRCh38
rs199920317 Chr.7: 37850432-37850432 on Build GRCh38
rs745571114 Chr.7: 37850653-37850653 on Build GRCh38
rs764519450 Chr.7: 37850395-37850395 on Build GRCh38
rs150406326 Chr.7: 37850599-37850599 on Build GRCh38
rs746723740 Chr.7: 37850617-37850617 on Build GRCh38
rs984892337 Chr.7: 37850340-37850340 on Build GRCh38
rs113388202 Chr.7: 37850620-37850620 on Build GRCh38
rs180763598 Chr.7: 37850708-37850708 on Build GRCh38
rs772405642 Chr.7: 37850671-37850671 on Build GRCh38
rs746669564 Chr.7: 37850451-37850451 on Build GRCh38
rs1002238773 Chr.7: 37850587-37850587 on Build GRCh38
rs2722371 Chr.7: 37850479-37850479 on Build GRCh38
rs769263780 Chr.7: 37850659-37850659 on Build GRCh38
rs746181533 Chr.7: 37850314-37850314 on Build GRCh38
rs376117068 Chr.7: 37850428-37850428 on Build GRCh38
rs199848335 Chr.7: 37850681-37850681 on Build GRCh38
rs767922928 Chr.7: 37850478-37850478 on Build GRCh38
rs374040658 Chr.7: 37850408-37850408 on Build GRCh38
rs759876007 Chr.7: 37850706-37850706 on Build GRCh38
rs780775214 Chr.7: 37850648-37850648 on Build GRCh38
rs768774717 Chr.7: 37850327-37850327 on Build GRCh38
rs1009304897 Chr.7: 37850506-37850506 on Build GRCh38
rs747880885 Chr.7: 37850352-37850352 on Build GRCh38
rs777610363 Chr.7: 37850610-37850610 on Build GRCh38
rs759219304 Chr.7: 37850420-37850430 on Build GRCh38
rs778952400 Chr.7: 37850434-37850434 on Build GRCh38
rs776350841 Chr.7: 37850461-37850461 on Build GRCh38
rs934943362 Chr.7: 37850397-37850397 on Build GRCh38
rs199576209 Chr.7: 37850283-37850283 on Build GRCh38
rs751780750 Chr.7: 37850496-37850496 on Build GRCh38
rs2598044 Chr.7: 37850714-37850714 on Build GRCh38

View Full Product Details