Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37864208-37864634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1450589 c.519_520insA p.R175fs*15 Chr.7: 37864412-37864413 on Build GRCh38
5460600 c.486G>A p.P162P Chr.7: 37864379-37864379 on Build GRCh38
3412017 c.486G>C p.P162P Chr.7: 37864379-37864379 on Build GRCh38
7324474 c.463T>C p.Y155H Chr.7: 37864356-37864356 on Build GRCh38
5780641 c.456G>A p.Q152Q Chr.7: 37864349-37864349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3841582 Chr.7: 37864464-37864466 on Build GRCh38
rs898091175 Chr.7: 37864560-37864560 on Build GRCh38
rs1025403702 Chr.7: 37864578-37864578 on Build GRCh38
rs968061811 Chr.7: 37864487-37864487 on Build GRCh38
rs927115962 Chr.7: 37864364-37864364 on Build GRCh38
rs183926718 Chr.7: 37864501-37864501 on Build GRCh38
rs756291276 Chr.7: 37864334-37864334 on Build GRCh38
rs773318395 Chr.7: 37864389-37864389 on Build GRCh38
rs572103524 Chr.7: 37864303-37864303 on Build GRCh38
rs760066403 Chr.7: 37864361-37864361 on Build GRCh38
rs763491394 Chr.7: 37864368-37864368 on Build GRCh38
rs200841100 Chr.7: 37864466-37864466 on Build GRCh38
rs981266761 Chr.7: 37864350-37864350 on Build GRCh38
rs771756299 Chr.7: 37864336-37864337 on Build GRCh38
rs398066793 Chr.7: 37864467-37864469 on Build GRCh38
rs371340468 Chr.7: 37864323-37864323 on Build GRCh38
rs3816417 Chr.7: 37864497-37864497 on Build GRCh38
rs916378575 Chr.7: 37864233-37864233 on Build GRCh38
rs771570015 Chr.7: 37864355-37864355 on Build GRCh38
rs117681493 Chr.7: 37864432-37864432 on Build GRCh38
rs778927416 Chr.7: 37864450-37864450 on Build GRCh38
rs919671662 Chr.7: 37864594-37864594 on Build GRCh38
rs767593869 Chr.7: 37864381-37864381 on Build GRCh38
rs1016453040 Chr.7: 37864559-37864559 on Build GRCh38
rs764429331 Chr.7: 37864372-37864372 on Build GRCh38
rs778922099 Chr.7: 37864345-37864345 on Build GRCh38
rs916430524 Chr.7: 37864584-37864584 on Build GRCh38
rs557740361 Chr.7: 37864605-37864605 on Build GRCh38
rs746713310 Chr.7: 37864473-37864473 on Build GRCh38
rs753236293 Chr.7: 37864298-37864298 on Build GRCh38
rs1006372929 Chr.7: 37864532-37864532 on Build GRCh38
rs971234156 Chr.7: 37864337-37864337 on Build GRCh38
rs750678432 Chr.7: 37864387-37864387 on Build GRCh38
rs970800694 Chr.7: 37864600-37864600 on Build GRCh38
rs973843063 Chr.7: 37864586-37864586 on Build GRCh38
rs755028493 Chr.7: 37864442-37864442 on Build GRCh38
rs781489577 Chr.7: 37864322-37864322 on Build GRCh38
rs1041787279 Chr.7: 37864403-37864403 on Build GRCh38
rs564369668 Chr.7: 37864464-37864464 on Build GRCh38
rs929638965 Chr.7: 37864263-37864263 on Build GRCh38
rs993484356 Chr.7: 37864566-37864566 on Build GRCh38
rs960661406 Chr.7: 37864517-37864517 on Build GRCh38
rs771044048 Chr.7: 37864479-37864479 on Build GRCh38
rs969198247 Chr.7: 37864585-37864585 on Build GRCh38
rs780087249 Chr.7: 37864338-37864338 on Build GRCh38
rs756237951 Chr.7: 37864392-37864392 on Build GRCh38
rs772924365 Chr.7: 37864358-37864358 on Build GRCh38
rs60479506 Chr.7: 37864244-37864244 on Build GRCh38
rs947804666 Chr.7: 37864237-37864237 on Build GRCh38
rs777244150 Chr.7: 37864469-37864469 on Build GRCh38
rs1050171544 Chr.7: 37864496-37864496 on Build GRCh38
rs200290963 Chr.7: 37864362-37864362 on Build GRCh38
rs747826248 Chr.7: 37864352-37864352 on Build GRCh38
rs962007132 Chr.7: 37864258-37864258 on Build GRCh38
rs200863038 Chr.7: 37864378-37864378 on Build GRCh38
rs749283857 Chr.7: 37864342-37864342 on Build GRCh38
rs199713986 Chr.7: 37864379-37864379 on Build GRCh38
rs3816418 Chr.7: 37864302-37864302 on Build GRCh38
rs747833460 Chr.7: 37864462-37864462 on Build GRCh38
rs780317892 Chr.7: 37864417-37864417 on Build GRCh38
rs990060500 Chr.7: 37864369-37864369 on Build GRCh38
rs3816419 Chr.7: 37864251-37864251 on Build GRCh38
rs567461763 Chr.7: 37864268-37864268 on Build GRCh38
rs758714010 Chr.7: 37864300-37864300 on Build GRCh38

View Full Product Details