Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127451394-127451887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
UniGene
Hs.495188
Gene Chromosome Location
Chr.9: 127451486-127503501 on Build GRCh38
Transcripts
NM_001005373.3 NM_001190723.2 NM_138361.5 XM_006717316.3 XM_017015283.1 XM_017015284.1 NM_000976.3

Gene Symbol
RPL12
Entrez Gene ID
6136
Gene Name
ribosomal protein L12
Gene Aliases
L12
UniGene
Hs.408054
Gene Chromosome Location
Chr.9: 127447674-127451432 on Build GRCh38
Transcripts
NM_001005373.3 NM_001190723.2 NM_138361.5 XM_006717316.3 XM_017015283.1 XM_017015284.1 NM_000976.3

SNP Detail

SNP ID Chromosome Location
rs928347797 Chr.9: 127451441-127451441 on Build GRCh38
rs376368787 Chr.9: 127451647-127451647 on Build GRCh38
rs928400616 Chr.9: 127451726-127451726 on Build GRCh38
rs1055710973 Chr.9: 127451715-127451715 on Build GRCh38
rs3808832 Chr.9: 127451550-127451550 on Build GRCh38
rs1043485301 Chr.9: 127451441-127451442 on Build GRCh38
rs920134002 Chr.9: 127451423-127451423 on Build GRCh38
rs900368744 Chr.9: 127451531-127451531 on Build GRCh38
rs970202043 Chr.9: 127451698-127451698 on Build GRCh38
rs1034541848 Chr.9: 127451466-127451466 on Build GRCh38
rs909545287 Chr.9: 127451523-127451523 on Build GRCh38
rs1016668719 Chr.9: 127451779-127451779 on Build GRCh38
rs543003414 Chr.9: 127451754-127451754 on Build GRCh38
rs577328150 Chr.9: 127451691-127451691 on Build GRCh38
rs983876913 Chr.9: 127451513-127451513 on Build GRCh38
rs995972401 Chr.9: 127451538-127451538 on Build GRCh38
rs868169253 Chr.9: 127451495-127451495 on Build GRCh38
rs746664653 Chr.9: 127451562-127451562 on Build GRCh38
rs1016034577 Chr.9: 127451582-127451582 on Build GRCh38
rs557383479 Chr.9: 127451452-127451452 on Build GRCh38
rs878946759 Chr.9: 127451628-127451628 on Build GRCh38
rs761364421 Chr.9: 127451552-127451552 on Build GRCh38
rs749626964 Chr.9: 127451785-127451787 on Build GRCh38
rs1015862144 Chr.9: 127451417-127451417 on Build GRCh38
rs946132017 Chr.9: 127451654-127451654 on Build GRCh38
rs1042999507 Chr.9: 127451767-127451767 on Build GRCh38
rs891445642 Chr.9: 127451484-127451484 on Build GRCh38
rs533863891 Chr.9: 127451476-127451476 on Build GRCh38
rs912926777 Chr.9: 127451759-127451759 on Build GRCh38
rs980215919 Chr.9: 127451716-127451716 on Build GRCh38
rs975344755 Chr.9: 127451529-127451529 on Build GRCh38
rs1014867805 Chr.9: 127451477-127451477 on Build GRCh38
rs556391869 Chr.9: 127451819-127451819 on Build GRCh38
rs886063453 Chr.9: 127451808-127451808 on Build GRCh38
rs988041470 Chr.9: 127451836-127451836 on Build GRCh38
rs1014504064 Chr.9: 127451732-127451732 on Build GRCh38
rs760403428 Chr.9: 127451659-127451659 on Build GRCh38
rs947063392 Chr.9: 127451763-127451763 on Build GRCh38
rs960086197 Chr.9: 127451747-127451747 on Build GRCh38
rs961960223 Chr.9: 127451420-127451420 on Build GRCh38
rs116498902 Chr.9: 127451542-127451542 on Build GRCh38
rs886063452 Chr.9: 127451789-127451791 on Build GRCh38
rs1028431081 Chr.9: 127451540-127451540 on Build GRCh38
rs571406961 Chr.9: 127451435-127451435 on Build GRCh38
rs767137099 Chr.9: 127451559-127451559 on Build GRCh38
rs975105024 Chr.9: 127451804-127451804 on Build GRCh38
rs921824982 Chr.9: 127451812-127451812 on Build GRCh38
rs775881443 Chr.9: 127451447-127451447 on Build GRCh38
rs553510399 Chr.9: 127451524-127451524 on Build GRCh38
rs886063451 Chr.9: 127451779-127451779 on Build GRCh38
rs1025795609 Chr.9: 127451503-127451503 on Build GRCh38
rs974333519 Chr.9: 127451421-127451421 on Build GRCh38
rs796478456 Chr.9: 127451545-127451545 on Build GRCh38
rs1029314259 Chr.9: 127451807-127451807 on Build GRCh38
rs913728563 Chr.9: 127451848-127451848 on Build GRCh38
rs886063450 Chr.9: 127451716-127451716 on Build GRCh38
rs1026264468 Chr.9: 127451758-127451758 on Build GRCh38
rs551599331 Chr.9: 127451429-127451429 on Build GRCh38
rs951243488 Chr.9: 127451506-127451506 on Build GRCh38
rs1001745572 Chr.9: 127451463-127451463 on Build GRCh38
rs955508111 Chr.9: 127451823-127451823 on Build GRCh38
rs904552931 Chr.9: 127451670-127451673 on Build GRCh38

View Full Product Details