Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148988255-148988845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC2
Entrez Gene ID
79628
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
UniGene
Hs.483784
Gene Chromosome Location
Chr.5: 148982150-149063174 on Build GRCh38
Transcripts
NM_024577.3

SNP Detail

SNP ID Chromosome Location
rs4543254 Chr.5: 148988691-148988691 on Build GRCh38
rs1013911527 Chr.5: 148988453-148988453 on Build GRCh38
rs752423955 Chr.5: 148988307-148988307 on Build GRCh38
rs996274902 Chr.5: 148988786-148988786 on Build GRCh38
rs139941418 Chr.5: 148988744-148988744 on Build GRCh38
rs148590947 Chr.5: 148988369-148988369 on Build GRCh38
rs539126111 Chr.5: 148988476-148988476 on Build GRCh38
rs983569714 Chr.5: 148988284-148988284 on Build GRCh38
rs995039312 Chr.5: 148988475-148988475 on Build GRCh38
rs1041713702 Chr.5: 148988546-148988546 on Build GRCh38
rs1053352608 Chr.5: 148988638-148988638 on Build GRCh38
rs770186542 Chr.5: 148988654-148988654 on Build GRCh38
rs539441280 Chr.5: 148988511-148988512 on Build GRCh38
rs765288122 Chr.5: 148988281-148988281 on Build GRCh38
rs769092559 Chr.5: 148988685-148988685 on Build GRCh38
rs935474535 Chr.5: 148988731-148988731 on Build GRCh38
rs1052633732 Chr.5: 148988743-148988743 on Build GRCh38
rs80295748 Chr.5: 148988381-148988381 on Build GRCh38
rs960182524 Chr.5: 148988635-148988635 on Build GRCh38
rs572365201 Chr.5: 148988555-148988555 on Build GRCh38
rs144423228 Chr.5: 148988333-148988333 on Build GRCh38
rs746011772 Chr.5: 148988592-148988592 on Build GRCh38
rs925820414 Chr.5: 148988562-148988562 on Build GRCh38
rs184120738 Chr.5: 148988561-148988561 on Build GRCh38
rs144778701 Chr.5: 148988573-148988573 on Build GRCh38
rs890627592 Chr.5: 148988721-148988721 on Build GRCh38
rs35392693 Chr.5: 148988803-148988804 on Build GRCh38
rs973372225 Chr.5: 148988657-148988657 on Build GRCh38
rs777791862 Chr.5: 148988358-148988358 on Build GRCh38
rs966113812 Chr.5: 148988540-148988540 on Build GRCh38
rs1049322278 Chr.5: 148988699-148988699 on Build GRCh38
rs932537049 Chr.5: 148988380-148988380 on Build GRCh38
rs1044071288 Chr.5: 148988663-148988663 on Build GRCh38
rs1035261876 Chr.5: 148988602-148988602 on Build GRCh38
rs950932536 Chr.5: 148988494-148988494 on Build GRCh38
rs575621364 Chr.5: 148988504-148988504 on Build GRCh38
rs978717116 Chr.5: 148988557-148988557 on Build GRCh38
rs572081330 Chr.5: 148988454-148988454 on Build GRCh38
rs377415024 Chr.5: 148988777-148988777 on Build GRCh38
rs147906062 Chr.5: 148988655-148988655 on Build GRCh38
rs1029057351 Chr.5: 148988805-148988805 on Build GRCh38
rs535181061 Chr.5: 148988441-148988441 on Build GRCh38
rs1047760995 Chr.5: 148988493-148988493 on Build GRCh38
rs983947009 Chr.5: 148988637-148988637 on Build GRCh38
rs1023240949 Chr.5: 148988773-148988773 on Build GRCh38
rs142027644 Chr.5: 148988430-148988430 on Build GRCh38
rs191600511 Chr.5: 148988542-148988542 on Build GRCh38
rs535652526 Chr.5: 148988527-148988527 on Build GRCh38
rs922207461 Chr.5: 148988671-148988671 on Build GRCh38
rs573762711 Chr.5: 148988550-148988550 on Build GRCh38
rs932150932 Chr.5: 148988526-148988526 on Build GRCh38
rs538758602 Chr.5: 148988820-148988820 on Build GRCh38
rs150690882 Chr.5: 148988450-148988450 on Build GRCh38
rs1019511165 Chr.5: 148988740-148988740 on Build GRCh38
rs549696782 Chr.5: 148988415-148988415 on Build GRCh38
rs749545774 Chr.5: 148988665-148988665 on Build GRCh38
rs75958253 Chr.5: 148988502-148988502 on Build GRCh38
rs954581706 Chr.5: 148988363-148988363 on Build GRCh38
rs116508083 Chr.5: 148988316-148988316 on Build GRCh38
rs981935495 Chr.5: 148988593-148988593 on Build GRCh38
rs914491112 Chr.5: 148988320-148988320 on Build GRCh38
rs1052873611 Chr.5: 148988446-148988446 on Build GRCh38
rs114694386 Chr.5: 148988445-148988445 on Build GRCh38
rs886657590 Chr.5: 148988702-148988702 on Build GRCh38
rs747266246 Chr.5: 148988462-148988462 on Build GRCh38
rs1010558241 Chr.5: 148988762-148988762 on Build GRCh38
rs1005090373 Chr.5: 148988709-148988709 on Build GRCh38
rs921221354 Chr.5: 148988378-148988378 on Build GRCh38
rs994438752 Chr.5: 148988696-148988696 on Build GRCh38
rs561967330 Chr.5: 148988543-148988543 on Build GRCh38
rs756518895 Chr.5: 148988640-148988640 on Build GRCh38

View Full Product Details