Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42744997-42745502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

SNP Detail

SNP ID Chromosome Location
rs538889580 Chr.15: 42745209-42745209 on Build GRCh38
rs928031530 Chr.15: 42745100-42745100 on Build GRCh38
rs910841140 Chr.15: 42745214-42745215 on Build GRCh38
rs886051161 Chr.15: 42745191-42745191 on Build GRCh38
rs540443679 Chr.15: 42745463-42745464 on Build GRCh38
rs768043177 Chr.15: 42745284-42745284 on Build GRCh38
rs968782952 Chr.15: 42745210-42745210 on Build GRCh38
rs536322846 Chr.15: 42745112-42745112 on Build GRCh38
rs78891022 Chr.15: 42745310-42745310 on Build GRCh38
rs930354225 Chr.15: 42745353-42745353 on Build GRCh38
rs544516599 Chr.15: 42745215-42745215 on Build GRCh38
rs902865683 Chr.15: 42745259-42745259 on Build GRCh38
rs991532320 Chr.15: 42745111-42745111 on Build GRCh38
rs575564250 Chr.15: 42745214-42745214 on Build GRCh38
rs1013224044 Chr.15: 42745046-42745046 on Build GRCh38
rs998531085 Chr.15: 42745283-42745283 on Build GRCh38
rs774535951 Chr.15: 42745197-42745197 on Build GRCh38
rs116853892 Chr.15: 42745212-42745212 on Build GRCh38
rs1022902264 Chr.15: 42745051-42745054 on Build GRCh38
rs1039978390 Chr.15: 42745240-42745240 on Build GRCh38
rs566913466 Chr.15: 42745207-42745207 on Build GRCh38
rs1047828037 Chr.15: 42745388-42745388 on Build GRCh38
rs769283772 Chr.15: 42745051-42745051 on Build GRCh38
rs138240929 Chr.15: 42745339-42745339 on Build GRCh38
rs892939920 Chr.15: 42745288-42745288 on Build GRCh38
rs887382606 Chr.15: 42745436-42745436 on Build GRCh38
rs1034983320 Chr.15: 42745350-42745350 on Build GRCh38
rs898909748 Chr.15: 42745337-42745337 on Build GRCh38
rs78237254 Chr.15: 42745117-42745117 on Build GRCh38
rs746273014 Chr.15: 42745181-42745181 on Build GRCh38
rs886051162 Chr.15: 42745288-42745288 on Build GRCh38
rs1027270353 Chr.15: 42745032-42745032 on Build GRCh38
rs181464499 Chr.15: 42745126-42745126 on Build GRCh38
rs987335300 Chr.15: 42745301-42745301 on Build GRCh38
rs962081721 Chr.15: 42745105-42745105 on Build GRCh38
rs186005779 Chr.15: 42745266-42745266 on Build GRCh38
rs886051160 Chr.15: 42745104-42745105 on Build GRCh38
rs920738016 Chr.15: 42745198-42745198 on Build GRCh38
rs887341765 Chr.15: 42745049-42745049 on Build GRCh38
rs75447904 Chr.15: 42745116-42745116 on Build GRCh38
rs960121592 Chr.15: 42745086-42745086 on Build GRCh38
rs748831261 Chr.15: 42745057-42745057 on Build GRCh38
rs16957120 Chr.15: 42745313-42745313 on Build GRCh38
rs142589086 Chr.15: 42745305-42745305 on Build GRCh38

View Full Product Details