Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95837890-95838479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR2
Entrez Gene ID
8898
Gene Name
myotubularin related protein 2
Gene Aliases
CMT4B, CMT4B1
UniGene
Hs.181326
Gene Chromosome Location
Chr.11: 95832880-95924207 on Build GRCh38
Transcripts
NM_001243571.1 NM_016156.5 NM_201278.2 NM_201281.2 XM_005274374.2 XM_005274375.2 XM_006718934.2 XM_006718935.2 XM_006718936.3 XM_011543058.2 XM_011543059.2 XM_017018517.1 XM_017018518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6716560 c.1585G>A p.G529R Chr.11: 95838102-95838102 on Build GRCh38
4529193 c.1377G>A p.E459E Chr.11: 95838094-95838094 on Build GRCh38
5361038 c.1314C>T p.D438D Chr.11: 95838157-95838157 on Build GRCh38
5987045 c.1549T>G p.F517V Chr.11: 95838138-95838138 on Build GRCh38
5361037 c.1530C>T p.D510D Chr.11: 95838157-95838157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756038624 Chr.11: 95838442-95838442 on Build GRCh38
rs769062222 Chr.11: 95838248-95838248 on Build GRCh38
rs546753588 Chr.11: 95838386-95838386 on Build GRCh38
rs781248324 Chr.11: 95838050-95838050 on Build GRCh38
rs754865965 Chr.11: 95838047-95838047 on Build GRCh38
rs777732110 Chr.11: 95838056-95838056 on Build GRCh38
rs544183621 Chr.11: 95838264-95838264 on Build GRCh38
rs1043658271 Chr.11: 95838018-95838018 on Build GRCh38
rs766105845 Chr.11: 95838173-95838173 on Build GRCh38
rs112327353 Chr.11: 95838199-95838199 on Build GRCh38
rs777869929 Chr.11: 95838213-95838213 on Build GRCh38
rs757409208 Chr.11: 95838214-95838214 on Build GRCh38
rs1040577400 Chr.11: 95838304-95838305 on Build GRCh38
rs368240951 Chr.11: 95838008-95838008 on Build GRCh38
rs147481973 Chr.11: 95838352-95838352 on Build GRCh38
rs779077686 Chr.11: 95838225-95838225 on Build GRCh38
rs772625760 Chr.11: 95838167-95838167 on Build GRCh38
rs920704287 Chr.11: 95838155-95838155 on Build GRCh38
rs771089932 Chr.11: 95838075-95838075 on Build GRCh38
rs556873132 Chr.11: 95837945-95837945 on Build GRCh38
rs942597739 Chr.11: 95838237-95838237 on Build GRCh38
rs536291683 Chr.11: 95838015-95838015 on Build GRCh38
rs752626893 Chr.11: 95838198-95838198 on Build GRCh38
rs1023773381 Chr.11: 95837949-95837949 on Build GRCh38
rs140720967 Chr.11: 95838238-95838238 on Build GRCh38
rs746006353 Chr.11: 95838211-95838212 on Build GRCh38
rs375602247 Chr.11: 95838231-95838231 on Build GRCh38
rs767138276 Chr.11: 95838207-95838208 on Build GRCh38
rs944506669 Chr.11: 95837981-95837981 on Build GRCh38
rs972154399 Chr.11: 95838062-95838062 on Build GRCh38
rs767480618 Chr.11: 95838194-95838194 on Build GRCh38
rs1001803015 Chr.11: 95838384-95838384 on Build GRCh38
rs892338175 Chr.11: 95838007-95838007 on Build GRCh38
rs944138728 Chr.11: 95838269-95838269 on Build GRCh38
rs752523757 Chr.11: 95838210-95838211 on Build GRCh38
rs370588663 Chr.11: 95838353-95838353 on Build GRCh38
rs775961979 Chr.11: 95838136-95838136 on Build GRCh38
rs562850541 Chr.11: 95838279-95838279 on Build GRCh38
rs762611200 Chr.11: 95838218-95838218 on Build GRCh38
rs144656779 Chr.11: 95838288-95838288 on Build GRCh38
rs371041744 Chr.11: 95838227-95838227 on Build GRCh38
rs61735578 Chr.11: 95838183-95838183 on Build GRCh38
rs183407455 Chr.11: 95838263-95838263 on Build GRCh38
rs543044953 Chr.11: 95838334-95838334 on Build GRCh38
rs935016020 Chr.11: 95838344-95838344 on Build GRCh38
rs754836790 Chr.11: 95838394-95838394 on Build GRCh38
rs903526034 Chr.11: 95838037-95838037 on Build GRCh38
rs777241424 Chr.11: 95838249-95838249 on Build GRCh38
rs746210433 Chr.11: 95838106-95838106 on Build GRCh38
rs756207944 Chr.11: 95838055-95838055 on Build GRCh38
rs551979088 Chr.11: 95837993-95837993 on Build GRCh38
rs767536319 Chr.11: 95838421-95838421 on Build GRCh38
rs1040222950 Chr.11: 95837988-95837988 on Build GRCh38
rs571514280 Chr.11: 95838395-95838395 on Build GRCh38
rs761100126 Chr.11: 95838156-95838156 on Build GRCh38
rs772490648 Chr.11: 95838124-95838124 on Build GRCh38
rs528582626 Chr.11: 95838365-95838365 on Build GRCh38
rs779201148 Chr.11: 95838104-95838104 on Build GRCh38
rs200892517 Chr.11: 95838185-95838185 on Build GRCh38
rs769280444 Chr.11: 95838164-95838164 on Build GRCh38
rs746144943 Chr.11: 95838226-95838226 on Build GRCh38
rs759161985 Chr.11: 95838166-95838166 on Build GRCh38
rs1001855356 Chr.11: 95838039-95838039 on Build GRCh38
rs954012752 Chr.11: 95838220-95838220 on Build GRCh38
rs749461263 Chr.11: 95838067-95838067 on Build GRCh38
rs913727280 Chr.11: 95838321-95838321 on Build GRCh38
rs117909641 Chr.11: 95838427-95838427 on Build GRCh38
rs758483872 Chr.11: 95838226-95838226 on Build GRCh38
rs370540902 Chr.11: 95838256-95838256 on Build GRCh38
rs762613780 Chr.11: 95838171-95838171 on Build GRCh38
rs1014132718 Chr.11: 95838135-95838135 on Build GRCh38
rs982775572 Chr.11: 95838028-95838028 on Build GRCh38
rs1015773638 Chr.11: 95838151-95838151 on Build GRCh38
rs747294899 Chr.11: 95838297-95838300 on Build GRCh38
rs765863380 Chr.11: 95838067-95838068 on Build GRCh38
rs751116352 Chr.11: 95838355-95838355 on Build GRCh38
rs113118572 Chr.11: 95838382-95838382 on Build GRCh38
rs773746206 Chr.11: 95838102-95838103 on Build GRCh38
rs749198789 Chr.11: 95838043-95838044 on Build GRCh38
rs1010631558 Chr.11: 95838372-95838372 on Build GRCh38
rs528180518 Chr.11: 95838413-95838413 on Build GRCh38
rs893530622 Chr.11: 95838360-95838360 on Build GRCh38

View Full Product Details