Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37857074-37857567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NME8
Entrez Gene ID
51314
Gene Name
NME/NM23 family member 8
Gene Aliases
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2
UniGene
Hs.723454
Gene Chromosome Location
Chr.7: 37848597-37900401 on Build GRCh38
Transcripts
NM_016616.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6727363 c.229C>T p.P77S Chr.7: 37857304-37857304 on Build GRCh38
5725530 c.235A>C p.R79R Chr.7: 37857310-37857310 on Build GRCh38
5407442 c.230C>T p.P77L Chr.7: 37857305-37857305 on Build GRCh38
3768357 c.258T>C p.F86F Chr.7: 37857333-37857333 on Build GRCh38
1488535 c.205G>C p.A69P Chr.7: 37857280-37857280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78352874 Chr.7: 37857099-37857099 on Build GRCh38
rs554401414 Chr.7: 37857152-37857152 on Build GRCh38
rs145204294 Chr.7: 37857213-37857213 on Build GRCh38
rs778271146 Chr.7: 37857390-37857390 on Build GRCh38
rs766933294 Chr.7: 37857335-37857335 on Build GRCh38
rs80215686 Chr.7: 37857222-37857222 on Build GRCh38
rs113718796 Chr.7: 37857209-37857209 on Build GRCh38
rs574943709 Chr.7: 37857102-37857102 on Build GRCh38
rs777289980 Chr.7: 37857324-37857324 on Build GRCh38
rs547826686 Chr.7: 37857339-37857339 on Build GRCh38
rs925705213 Chr.7: 37857234-37857234 on Build GRCh38
rs961245558 Chr.7: 37857188-37857188 on Build GRCh38
rs762892925 Chr.7: 37857230-37857230 on Build GRCh38
rs757288576 Chr.7: 37857251-37857251 on Build GRCh38
rs541494396 Chr.7: 37857299-37857299 on Build GRCh38
rs913026276 Chr.7: 37857532-37857532 on Build GRCh38
rs890541626 Chr.7: 37857530-37857530 on Build GRCh38
rs892022358 Chr.7: 37857126-37857126 on Build GRCh38
rs763736855 Chr.7: 37857101-37857101 on Build GRCh38
rs887838219 Chr.7: 37857377-37857377 on Build GRCh38
rs944902538 Chr.7: 37857320-37857320 on Build GRCh38
rs202034687 Chr.7: 37857328-37857328 on Build GRCh38
rs1018921088 Chr.7: 37857134-37857134 on Build GRCh38
rs150983980 Chr.7: 37857386-37857386 on Build GRCh38
rs942215199 Chr.7: 37857116-37857116 on Build GRCh38
rs781770917 Chr.7: 37857397-37857397 on Build GRCh38
rs750395044 Chr.7: 37857365-37857365 on Build GRCh38
rs755910817 Chr.7: 37857275-37857275 on Build GRCh38
rs756851927 Chr.7: 37857184-37857184 on Build GRCh38
rs925624411 Chr.7: 37857435-37857435 on Build GRCh38
rs527437340 Chr.7: 37857378-37857378 on Build GRCh38
rs140475619 Chr.7: 37857502-37857502 on Build GRCh38
rs943373800 Chr.7: 37857535-37857535 on Build GRCh38
rs540932392 Chr.7: 37857135-37857135 on Build GRCh38
rs758846066 Chr.7: 37857318-37857319 on Build GRCh38
rs981830782 Chr.7: 37857355-37857355 on Build GRCh38
rs540579794 Chr.7: 37857146-37857146 on Build GRCh38
rs183351573 Chr.7: 37857192-37857192 on Build GRCh38
rs1038333726 Chr.7: 37857418-37857418 on Build GRCh38
rs763278466 Chr.7: 37857336-37857336 on Build GRCh38
rs774550075 Chr.7: 37857345-37857345 on Build GRCh38
rs750163886 Chr.7: 37857273-37857273 on Build GRCh38
rs576813254 Chr.7: 37857163-37857163 on Build GRCh38
rs369809235 Chr.7: 37857367-37857367 on Build GRCh38
rs748610824 Chr.7: 37857526-37857526 on Build GRCh38
rs751423260 Chr.7: 37857247-37857247 on Build GRCh38
rs373963901 Chr.7: 37857370-37857370 on Build GRCh38
rs996218149 Chr.7: 37857148-37857148 on Build GRCh38
rs558885140 Chr.7: 37857239-37857239 on Build GRCh38
rs775554270 Chr.7: 37857333-37857333 on Build GRCh38
rs919434453 Chr.7: 37857457-37857457 on Build GRCh38
rs961190679 Chr.7: 37857388-37857388 on Build GRCh38
rs771622383 Chr.7: 37857319-37857319 on Build GRCh38
rs952250781 Chr.7: 37857159-37857159 on Build GRCh38
rs746168002 Chr.7: 37857325-37857325 on Build GRCh38
rs142570057 Chr.7: 37857301-37857301 on Build GRCh38
rs113108115 Chr.7: 37857158-37857158 on Build GRCh38
rs969969364 Chr.7: 37857413-37857413 on Build GRCh38
rs1024161326 Chr.7: 37857398-37857398 on Build GRCh38
rs779645085 Chr.7: 37857289-37857289 on Build GRCh38
rs3213976 Chr.7: 37857294-37857294 on Build GRCh38
rs979771654 Chr.7: 37857423-37857423 on Build GRCh38
rs929425377 Chr.7: 37857473-37857473 on Build GRCh38
rs747465574 Chr.7: 37857314-37857314 on Build GRCh38
rs767379954 Chr.7: 37857357-37857357 on Build GRCh38
rs377333583 Chr.7: 37857182-37857182 on Build GRCh38
rs75446846 Chr.7: 37857220-37857220 on Build GRCh38
rs1009165633 Chr.7: 37857127-37857127 on Build GRCh38
rs569710064 Chr.7: 37857454-37857454 on Build GRCh38
rs767289819 Chr.7: 37857253-37857253 on Build GRCh38
rs377202353 Chr.7: 37857354-37857354 on Build GRCh38
rs12673299 Chr.7: 37857510-37857510 on Build GRCh38

View Full Product Details