Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27367069-27367302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6333235 c.999C>A p.I333I Chr.2: 27367088-27367088 on Build GRCh38
6091445 c.977G>T p.R326L Chr.2: 27367170-27367170 on Build GRCh38
4936978 c.990G>A p.E330E Chr.2: 27367157-27367157 on Build GRCh38
7316781 c.988G>C p.E330Q Chr.2: 27367159-27367159 on Build GRCh38
4820146 c.955G>C p.E319Q Chr.2: 27367192-27367192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763515698 Chr.2: 27367264-27367264 on Build GRCh38
rs776661452 Chr.2: 27367152-27367152 on Build GRCh38
rs377339042 Chr.2: 27367180-27367180 on Build GRCh38
rs746687079 Chr.2: 27367106-27367106 on Build GRCh38
rs773304003 Chr.2: 27367133-27367133 on Build GRCh38
rs538015161 Chr.2: 27367234-27367234 on Build GRCh38
rs745692536 Chr.2: 27367230-27367230 on Build GRCh38
rs769906014 Chr.2: 27367262-27367262 on Build GRCh38
rs774091341 Chr.2: 27367229-27367229 on Build GRCh38
rs780633076 Chr.2: 27367115-27367115 on Build GRCh38
rs373520608 Chr.2: 27367157-27367157 on Build GRCh38
rs78599355 Chr.2: 27367171-27367171 on Build GRCh38
rs780909643 Chr.2: 27367182-27367182 on Build GRCh38
rs750965977 Chr.2: 27367161-27367161 on Build GRCh38
rs755715511 Chr.2: 27367186-27367186 on Build GRCh38
rs760874790 Chr.2: 27367147-27367147 on Build GRCh38
rs200893372 Chr.2: 27367222-27367222 on Build GRCh38
rs200954953 Chr.2: 27367126-27367126 on Build GRCh38
rs560532019 Chr.2: 27367149-27367149 on Build GRCh38
rs761908506 Chr.2: 27367154-27367154 on Build GRCh38
rs770898806 Chr.2: 27367138-27367138 on Build GRCh38
rs773264189 Chr.2: 27367263-27367263 on Build GRCh38
rs549609482 Chr.2: 27367213-27367213 on Build GRCh38
rs749208137 Chr.2: 27367136-27367136 on Build GRCh38
rs781536399 Chr.2: 27367185-27367185 on Build GRCh38
rs184607650 Chr.2: 27367140-27367140 on Build GRCh38
rs758877324 Chr.2: 27367166-27367166 on Build GRCh38
rs754642030 Chr.2: 27367107-27367107 on Build GRCh38
rs769371305 Chr.2: 27367132-27367132 on Build GRCh38
rs761988390 Chr.2: 27367240-27367240 on Build GRCh38
rs370668762 Chr.2: 27367237-27367237 on Build GRCh38
rs770990507 Chr.2: 27367223-27367223 on Build GRCh38

View Full Product Details