Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27367328-27367593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4648585 c.860G>A p.R287H Chr.2: 27367542-27367542 on Build GRCh38
6569130 c.924C>T p.G308G Chr.2: 27367478-27367478 on Build GRCh38
2939888 c.826G>A p.A276T Chr.2: 27367516-27367516 on Build GRCh38
1407432 c.862G>T p.G288C Chr.2: 27367480-27367480 on Build GRCh38
1193374 c.865A>C p.S289R Chr.2: 27367477-27367477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767982696 Chr.2: 27367410-27367410 on Build GRCh38
rs1036710491 Chr.2: 27367361-27367361 on Build GRCh38
rs571958406 Chr.2: 27367375-27367375 on Build GRCh38
rs751447134 Chr.2: 27367568-27367568 on Build GRCh38
rs774281750 Chr.2: 27367513-27367513 on Build GRCh38
rs554814067 Chr.2: 27367388-27367388 on Build GRCh38
rs577147537 Chr.2: 27367477-27367477 on Build GRCh38
rs749471127 Chr.2: 27367504-27367504 on Build GRCh38
rs898468416 Chr.2: 27367478-27367478 on Build GRCh38
rs912337423 Chr.2: 27367359-27367359 on Build GRCh38
rs754707878 Chr.2: 27367479-27367479 on Build GRCh38
rs190848009 Chr.2: 27367473-27367473 on Build GRCh38
rs180670782 Chr.2: 27367376-27367376 on Build GRCh38
rs939789780 Chr.2: 27367361-27367362 on Build GRCh38
rs957399650 Chr.2: 27367539-27367539 on Build GRCh38
rs775042965 Chr.2: 27367536-27367537 on Build GRCh38
rs764856179 Chr.2: 27367544-27367544 on Build GRCh38
rs553632153 Chr.2: 27367371-27367371 on Build GRCh38
rs149782530 Chr.2: 27367517-27367517 on Build GRCh38
rs375252124 Chr.2: 27367449-27367449 on Build GRCh38
rs758251555 Chr.2: 27367467-27367467 on Build GRCh38
rs368372393 Chr.2: 27367470-27367470 on Build GRCh38
rs764226813 Chr.2: 27367370-27367370 on Build GRCh38
rs753108196 Chr.2: 27367426-27367426 on Build GRCh38
rs374767999 Chr.2: 27367402-27367402 on Build GRCh38
rs761353843 Chr.2: 27367538-27367538 on Build GRCh38
rs761145823 Chr.2: 27367529-27367529 on Build GRCh38
rs538217203 Chr.2: 27367496-27367496 on Build GRCh38
rs779887864 Chr.2: 27367468-27367468 on Build GRCh38
rs763088766 Chr.2: 27367462-27367462 on Build GRCh38
rs777992006 Chr.2: 27367501-27367501 on Build GRCh38
rs889851700 Chr.2: 27367543-27367543 on Build GRCh38
rs182331835 Chr.2: 27367530-27367530 on Build GRCh38
rs543764951 Chr.2: 27367409-27367409 on Build GRCh38
rs753494246 Chr.2: 27367430-27367430 on Build GRCh38
rs771082359 Chr.2: 27367505-27367505 on Build GRCh38
rs143926434 Chr.2: 27367551-27367551 on Build GRCh38
rs766314579 Chr.2: 27367566-27367566 on Build GRCh38
rs1028342862 Chr.2: 27367531-27367531 on Build GRCh38
rs770865097 Chr.2: 27367407-27367407 on Build GRCh38
rs185104957 Chr.2: 27367408-27367408 on Build GRCh38
rs767850637 Chr.2: 27367474-27367474 on Build GRCh38
rs1013108994 Chr.2: 27367393-27367393 on Build GRCh38
rs1056952345 Chr.2: 27367387-27367387 on Build GRCh38
rs113994031 Chr.2: 27367536-27367536 on Build GRCh38
rs544086994 Chr.2: 27367401-27367402 on Build GRCh38
rs947042721 Chr.2: 27367358-27367358 on Build GRCh38
rs898173808 Chr.2: 27367363-27367363 on Build GRCh38
rs756826232 Chr.2: 27367434-27367434 on Build GRCh38
rs763853754 Chr.2: 27367429-27367429 on Build GRCh38
rs549126918 Chr.2: 27367562-27367562 on Build GRCh38
rs777016486 Chr.2: 27367428-27367428 on Build GRCh38

View Full Product Details