Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18121635-18121884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHLRC1
Entrez Gene ID
378884
Gene Name
NHL repeat containing E3 ubiquitin protein ligase 1
Gene Aliases
EPM2A, EPM2B, MALIN, bA204B7.2
UniGene
Hs.348351
Gene Chromosome Location
Chr.6: 18120487-18122620 on Build GRCh38
Transcripts
NM_198586.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4506572 c.723C>T p.L241L Chr.6: 18121884-18121884 on Build GRCh38
6455892 c.869G>T p.C290F Chr.6: 18121738-18121738 on Build GRCh38
6513781 c.794G>T p.R265L Chr.6: 18121813-18121813 on Build GRCh38
2856205 c.904C>T p.Q302* Chr.6: 18121703-18121703 on Build GRCh38
405166 c.958_959insA p.I320fs*9 Chr.6: 18121648-18121649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780828215 Chr.6: 18121809-18121809 on Build GRCh38
rs778928979 Chr.6: 18121826-18121826 on Build GRCh38
rs774397860 Chr.6: 18121657-18121657 on Build GRCh38
rs768626564 Chr.6: 18121824-18121824 on Build GRCh38
rs757858146 Chr.6: 18121702-18121702 on Build GRCh38
rs749287999 Chr.6: 18121820-18121820 on Build GRCh38
rs201855901 Chr.6: 18121742-18121742 on Build GRCh38
rs773378991 Chr.6: 18121831-18121831 on Build GRCh38
rs568213488 Chr.6: 18121787-18121787 on Build GRCh38
rs981579921 Chr.6: 18121822-18121822 on Build GRCh38
rs750788641 Chr.6: 18121680-18121680 on Build GRCh38
rs750055958 Chr.6: 18121865-18121865 on Build GRCh38
rs746676778 Chr.6: 18121713-18121713 on Build GRCh38
rs121917875 Chr.6: 18121814-18121814 on Build GRCh38
rs377025668 Chr.6: 18121834-18121834 on Build GRCh38
rs776953775 Chr.6: 18121839-18121839 on Build GRCh38
rs777509826 Chr.6: 18121711-18121711 on Build GRCh38
rs780904929 Chr.6: 18121728-18121728 on Build GRCh38
rs796052756 Chr.6: 18121816-18121816 on Build GRCh38
rs755474952 Chr.6: 18121799-18121799 on Build GRCh38
rs756437109 Chr.6: 18121759-18121759 on Build GRCh38
rs371072212 Chr.6: 18121852-18121852 on Build GRCh38
rs779965723 Chr.6: 18121819-18121819 on Build GRCh38
rs756925518 Chr.6: 18121808-18121808 on Build GRCh38
rs772199816 Chr.6: 18121829-18121829 on Build GRCh38
rs1032004459 Chr.6: 18121738-18121738 on Build GRCh38
rs374327275 Chr.6: 18121856-18121856 on Build GRCh38
rs554356127 Chr.6: 18121751-18121751 on Build GRCh38
rs367906378 Chr.6: 18121867-18121867 on Build GRCh38
rs137852859 Chr.6: 18121684-18121684 on Build GRCh38
rs571973789 Chr.6: 18121866-18121866 on Build GRCh38
rs556216487 Chr.6: 18121776-18121776 on Build GRCh38
rs527299943 Chr.6: 18121674-18121674 on Build GRCh38
rs140066702 Chr.6: 18121770-18121770 on Build GRCh38
rs766807637 Chr.6: 18121689-18121689 on Build GRCh38
rs756909945 Chr.6: 18121727-18121727 on Build GRCh38
rs979143914 Chr.6: 18121678-18121678 on Build GRCh38
rs761062592 Chr.6: 18121833-18121833 on Build GRCh38
rs752220824 Chr.6: 18121694-18121694 on Build GRCh38
rs745652384 Chr.6: 18121732-18121732 on Build GRCh38
rs994146351 Chr.6: 18121688-18121688 on Build GRCh38
rs762064606 Chr.6: 18121664-18121664 on Build GRCh38
rs756683363 Chr.6: 18121685-18121685 on Build GRCh38
rs767777371 Chr.6: 18121769-18121769 on Build GRCh38
rs760017380 Chr.6: 18121849-18121849 on Build GRCh38
rs879745047 Chr.6: 18121825-18121825 on Build GRCh38
rs767300701 Chr.6: 18121861-18121861 on Build GRCh38
rs762001530 Chr.6: 18121764-18121764 on Build GRCh38
rs553814041 Chr.6: 18121813-18121813 on Build GRCh38
rs140164729 Chr.6: 18121802-18121802 on Build GRCh38
rs768659412 Chr.6: 18121743-18121743 on Build GRCh38
rs535618322 Chr.6: 18121806-18121806 on Build GRCh38
rs144043056 Chr.6: 18121828-18121828 on Build GRCh38
rs375475285 Chr.6: 18121752-18121752 on Build GRCh38
rs1029669440 Chr.6: 18121700-18121700 on Build GRCh38
rs200214191 Chr.6: 18121733-18121733 on Build GRCh38

View Full Product Details