Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35075352-35075611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCG
Entrez Gene ID
2189
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
UniGene
Hs.591084
Gene Chromosome Location
Chr.9: 35073838-35080016 on Build GRCh38
Transcripts
NM_004629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6295281 c.1341C>A p.Y447* Chr.9: 35075557-35075557 on Build GRCh38
1108587 c.1351T>C p.W451R Chr.9: 35075547-35075547 on Build GRCh38
6052524 c.1291A>G p.M431V Chr.9: 35075607-35075607 on Build GRCh38
6338085 c.1320A>G p.K440K Chr.9: 35075578-35075578 on Build GRCh38
1108586 c.1402G>A p.A468T Chr.9: 35075496-35075496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778986343 Chr.9: 35075557-35075557 on Build GRCh38
rs187315404 Chr.9: 35075456-35075456 on Build GRCh38
rs757442131 Chr.9: 35075552-35075552 on Build GRCh38
rs756155858 Chr.9: 35075435-35075435 on Build GRCh38
rs756069860 Chr.9: 35075544-35075544 on Build GRCh38
rs776133588 Chr.9: 35075475-35075475 on Build GRCh38
rs769402230 Chr.9: 35075490-35075490 on Build GRCh38
rs767385265 Chr.9: 35075533-35075533 on Build GRCh38
rs764116943 Chr.9: 35075545-35075545 on Build GRCh38
rs191508370 Chr.9: 35075570-35075570 on Build GRCh38
rs777880901 Chr.9: 35075441-35075441 on Build GRCh38
rs201070684 Chr.9: 35075453-35075453 on Build GRCh38
rs897901007 Chr.9: 35075573-35075573 on Build GRCh38
rs376961485 Chr.9: 35075444-35075444 on Build GRCh38
rs200456325 Chr.9: 35075566-35075566 on Build GRCh38
rs746164729 Chr.9: 35075558-35075558 on Build GRCh38
rs148808709 Chr.9: 35075531-35075531 on Build GRCh38
rs948298405 Chr.9: 35075420-35075420 on Build GRCh38
rs766157946 Chr.9: 35075468-35075468 on Build GRCh38
rs762631021 Chr.9: 35075499-35075499 on Build GRCh38
rs587778345 Chr.9: 35075532-35075532 on Build GRCh38
rs989531823 Chr.9: 35075396-35075396 on Build GRCh38
rs751287895 Chr.9: 35075526-35075526 on Build GRCh38
rs772794286 Chr.9: 35075587-35075587 on Build GRCh38
rs980822010 Chr.9: 35075470-35075470 on Build GRCh38
rs754889211 Chr.9: 35075417-35075417 on Build GRCh38
rs200078931 Chr.9: 35075419-35075419 on Build GRCh38
rs758656355 Chr.9: 35075560-35075560 on Build GRCh38
rs906662045 Chr.9: 35075485-35075485 on Build GRCh38
rs772542885 Chr.9: 35075457-35075457 on Build GRCh38
rs927127258 Chr.9: 35075478-35075478 on Build GRCh38
rs772883935 Chr.9: 35075492-35075492 on Build GRCh38
rs754635557 Chr.9: 35075576-35075576 on Build GRCh38
rs752700756 Chr.9: 35075434-35075434 on Build GRCh38
rs769293655 Chr.9: 35075578-35075578 on Build GRCh38
rs753917194 Chr.9: 35075548-35075548 on Build GRCh38
rs780612204 Chr.9: 35075563-35075563 on Build GRCh38
rs765965051 Chr.9: 35075512-35075512 on Build GRCh38
rs144369518 Chr.9: 35075476-35075476 on Build GRCh38
rs779268127 Chr.9: 35075454-35075454 on Build GRCh38

View Full Product Details