Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16988261-16988514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
UniGene
Hs.128866
Gene Chromosome Location
Chr.1: 16985958-17011972 on Build GRCh38
Transcripts
NM_001141973.2 NM_001141974.2 NM_022089.3 XM_005245810.1 XM_005245811.1 XM_005245812.1 XM_005245815.1 XM_006710512.1 XM_006710513.1 XM_011541128.1 XM_011541129.1 XM_017000844.1 XM_017000845.1 XM_017000846.1 XM_017000847.1 XM_017000848.1 XM_017000849.1 XM_017000850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6482900 c.2635G>T p.G879C Chr.1: 16988449-16988449 on Build GRCh38
4787841 c.2685G>T p.L895L Chr.1: 16988399-16988399 on Build GRCh38
6607274 c.2519G>A p.G840E Chr.1: 16988433-16988433 on Build GRCh38
1295498 c.2630G>C p.G877A Chr.1: 16988454-16988454 on Build GRCh38
4483261 c.2556C>T p.S852S Chr.1: 16988396-16988396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985370872 Chr.1: 16988448-16988448 on Build GRCh38
rs796448884 Chr.1: 16988291-16988293 on Build GRCh38
rs763069105 Chr.1: 16988361-16988361 on Build GRCh38
rs756683120 Chr.1: 16988409-16988409 on Build GRCh38
rs775892776 Chr.1: 16988463-16988463 on Build GRCh38
rs779656439 Chr.1: 16988303-16988303 on Build GRCh38
rs190323680 Chr.1: 16988421-16988421 on Build GRCh38
rs757945614 Chr.1: 16988469-16988469 on Build GRCh38
rs749964479 Chr.1: 16988420-16988420 on Build GRCh38
rs146234171 Chr.1: 16988342-16988342 on Build GRCh38
rs755414129 Chr.1: 16988281-16988281 on Build GRCh38
rs745504274 Chr.1: 16988299-16988299 on Build GRCh38
rs199994961 Chr.1: 16988465-16988465 on Build GRCh38
rs376189607 Chr.1: 16988483-16988483 on Build GRCh38
rs767865035 Chr.1: 16988461-16988461 on Build GRCh38
rs766632717 Chr.1: 16988362-16988362 on Build GRCh38
rs567287489 Chr.1: 16988444-16988444 on Build GRCh38
rs200702121 Chr.1: 16988284-16988284 on Build GRCh38
rs138437271 Chr.1: 16988399-16988399 on Build GRCh38
rs746804111 Chr.1: 16988304-16988304 on Build GRCh38
rs371975975 Chr.1: 16988429-16988429 on Build GRCh38
rs200504984 Chr.1: 16988467-16988467 on Build GRCh38
rs144701072 Chr.1: 16988455-16988455 on Build GRCh38
rs924044483 Chr.1: 16988484-16988484 on Build GRCh38
rs769961390 Chr.1: 16988356-16988356 on Build GRCh38
rs79724242 Chr.1: 16988360-16988360 on Build GRCh38
rs9435662 Chr.1: 16988447-16988447 on Build GRCh38
rs752015774 Chr.1: 16988278-16988278 on Build GRCh38
rs749859324 Chr.1: 16988468-16988468 on Build GRCh38
rs372361807 Chr.1: 16988339-16988339 on Build GRCh38
rs771014262 Chr.1: 16988449-16988449 on Build GRCh38
rs561561604 Chr.1: 16988456-16988456 on Build GRCh38
rs145455452 Chr.1: 16988375-16988375 on Build GRCh38
rs764639673 Chr.1: 16988411-16988411 on Build GRCh38
rs767921922 Chr.1: 16988402-16988402 on Build GRCh38
rs754741241 Chr.1: 16988494-16988494 on Build GRCh38
rs368772418 Chr.1: 16988470-16988470 on Build GRCh38
rs746749073 Chr.1: 16988424-16988424 on Build GRCh38
rs768674400 Chr.1: 16988338-16988338 on Build GRCh38
rs769823261 Chr.1: 16988433-16988433 on Build GRCh38
rs748091717 Chr.1: 16988432-16988432 on Build GRCh38
rs530039964 Chr.1: 16988332-16988332 on Build GRCh38
rs560897844 Chr.1: 16988301-16988301 on Build GRCh38
rs141447403 Chr.1: 16988293-16988293 on Build GRCh38
rs148336170 Chr.1: 16988291-16988291 on Build GRCh38
rs376348581 Chr.1: 16988403-16988403 on Build GRCh38
rs754796671 Chr.1: 16988426-16988426 on Build GRCh38
rs556418739 Chr.1: 16988450-16988450 on Build GRCh38
rs201437622 Chr.1: 16988296-16988296 on Build GRCh38

View Full Product Details