Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21302413-21302686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPGRIP1
Entrez Gene ID
57096
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
UniGene
Hs.126035
Gene Chromosome Location
Chr.14: 21287977-21351316 on Build GRCh38
Transcripts
NM_020366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4939972 c.503G>A p.R168K Chr.14: 21302500-21302500 on Build GRCh38
6045954 c.517G>T p.A173S Chr.14: 21302514-21302514 on Build GRCh38
6015442 c.527C>T p.S176L Chr.14: 21302524-21302524 on Build GRCh38
5538360 c.583G>A p.E195K Chr.14: 21302580-21302580 on Build GRCh38
3999219 c.574A>G p.K192E Chr.14: 21302571-21302571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757555537 Chr.14: 21302501-21302501 on Build GRCh38
rs17792599 Chr.14: 21302522-21302522 on Build GRCh38
rs749376395 Chr.14: 21302485-21302485 on Build GRCh38
rs199840691 Chr.14: 21302490-21302490 on Build GRCh38
rs370751257 Chr.14: 21302566-21302566 on Build GRCh38
rs747681631 Chr.14: 21302617-21302617 on Build GRCh38
rs1011857075 Chr.14: 21302524-21302524 on Build GRCh38
rs575192944 Chr.14: 21302507-21302507 on Build GRCh38
rs754595472 Chr.14: 21302615-21302615 on Build GRCh38
rs900955113 Chr.14: 21302546-21302546 on Build GRCh38
rs757653708 Chr.14: 21302578-21302578 on Build GRCh38
rs780733881 Chr.14: 21302518-21302518 on Build GRCh38
rs1046884011 Chr.14: 21302458-21302458 on Build GRCh38
rs1010719651 Chr.14: 21302625-21302625 on Build GRCh38
rs1055094187 Chr.14: 21302440-21302440 on Build GRCh38
rs376490441 Chr.14: 21302456-21302456 on Build GRCh38
rs369739531 Chr.14: 21302441-21302441 on Build GRCh38
rs753192367 Chr.14: 21302461-21302461 on Build GRCh38
rs1028039987 Chr.14: 21302586-21302586 on Build GRCh38
rs772938351 Chr.14: 21302527-21302527 on Build GRCh38
rs774735061 Chr.14: 21302540-21302540 on Build GRCh38
rs758926176 Chr.14: 21302568-21302568 on Build GRCh38
rs375351235 Chr.14: 21302525-21302525 on Build GRCh38
rs751765640 Chr.14: 21302451-21302451 on Build GRCh38
rs746627506 Chr.14: 21302513-21302513 on Build GRCh38
rs1002507485 Chr.14: 21302495-21302495 on Build GRCh38
rs550069974 Chr.14: 21302658-21302658 on Build GRCh38
rs200219549 Chr.14: 21302562-21302562 on Build GRCh38
rs61751265 Chr.14: 21302508-21302508 on Build GRCh38
rs767654540 Chr.14: 21302459-21302459 on Build GRCh38
rs113696227 Chr.14: 21302442-21302442 on Build GRCh38
rs751265096 Chr.14: 21302612-21302612 on Build GRCh38
rs764430814 Chr.14: 21302489-21302489 on Build GRCh38
rs754227744 Chr.14: 21302576-21302576 on Build GRCh38
rs751501153 Chr.14: 21302531-21302531 on Build GRCh38
rs6571751 Chr.14: 21302571-21302571 on Build GRCh38
rs754509080 Chr.14: 21302516-21302516 on Build GRCh38
rs574462207 Chr.14: 21302561-21302561 on Build GRCh38
rs1021312882 Chr.14: 21302638-21302638 on Build GRCh38
rs765540813 Chr.14: 21302609-21302609 on Build GRCh38
rs986513135 Chr.14: 21302602-21302602 on Build GRCh38
rs759808929 Chr.14: 21302545-21302545 on Build GRCh38
rs186266220 Chr.14: 21302539-21302539 on Build GRCh38

View Full Product Details