Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85981624-85981879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
UniGene
Hs.163893
Gene Chromosome Location
Chr.11: 85957171-86069881 on Build GRCh38
Transcripts
NM_001008660.2 NM_001206946.1 NM_001206947.1 NM_007166.3 XM_005274322.2 XM_005274323.2 XM_005274324.2 XM_005274325.2 XM_005274326.2 XM_005274327.2 XM_005274328.2 XM_005274329.3 XM_005274330.2 XM_005274331.2 XM_005274332.2 XM_005274333.2 XM_005274334.2 XM_005274335.2 XM_005274336.2 XM_005274337.2 XM_005274340.2 XM_006718699.2 XM_006718700.2 XM_006718701.3 XM_011545293.1 XM_017018381.1 XM_017018382.1 XM_017018383.1 XM_017018384.1 XM_017018385.1 XM_017018386.1 XM_017018387.1 XM_017018388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
351946 c.1666_1667GG>TA p.G556>? Chr.11: 85981757-85981758 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773884885 Chr.11: 85981769-85981769 on Build GRCh38
rs777268904 Chr.11: 85981702-85981705 on Build GRCh38
rs374145283 Chr.11: 85981816-85981816 on Build GRCh38
rs751763097 Chr.11: 85981813-85981813 on Build GRCh38
rs199605178 Chr.11: 85981852-85981852 on Build GRCh38
rs548939496 Chr.11: 85981853-85981853 on Build GRCh38
rs146684583 Chr.11: 85981796-85981796 on Build GRCh38
rs769899988 Chr.11: 85981762-85981762 on Build GRCh38
rs760028064 Chr.11: 85981797-85981797 on Build GRCh38
rs752993404 Chr.11: 85981804-85981804 on Build GRCh38
rs376324400 Chr.11: 85981839-85981839 on Build GRCh38
rs757538611 Chr.11: 85981814-85981814 on Build GRCh38
rs781144984 Chr.11: 85981745-85981745 on Build GRCh38
rs774013052 Chr.11: 85981842-85981842 on Build GRCh38
rs528175438 Chr.11: 85981765-85981765 on Build GRCh38
rs768284766 Chr.11: 85981767-85981767 on Build GRCh38
rs755822531 Chr.11: 85981785-85981785 on Build GRCh38
rs376390825 Chr.11: 85981714-85981714 on Build GRCh38
rs199763847 Chr.11: 85981784-85981784 on Build GRCh38
rs775411805 Chr.11: 85981764-85981764 on Build GRCh38
rs758775460 Chr.11: 85981805-85981805 on Build GRCh38
rs560190649 Chr.11: 85981660-85981660 on Build GRCh38
rs747052620 Chr.11: 85981729-85981729 on Build GRCh38
rs749010123 Chr.11: 85981729-85981733 on Build GRCh38
rs1047294021 Chr.11: 85981730-85981730 on Build GRCh38
rs780141981 Chr.11: 85981825-85981825 on Build GRCh38
rs747737013 Chr.11: 85981846-85981846 on Build GRCh38
rs200942143 Chr.11: 85981713-85981713 on Build GRCh38
rs746967111 Chr.11: 85981711-85981711 on Build GRCh38
rs745766988 Chr.11: 85981753-85981753 on Build GRCh38
rs572113237 Chr.11: 85981763-85981763 on Build GRCh38
rs756115600 Chr.11: 85981819-85981819 on Build GRCh38
rs370774354 Chr.11: 85981770-85981770 on Build GRCh38
rs1011974907 Chr.11: 85981850-85981850 on Build GRCh38
rs777863157 Chr.11: 85981728-85981728 on Build GRCh38
rs746002038 Chr.11: 85981818-85981818 on Build GRCh38
rs764330343 Chr.11: 85981807-85981807 on Build GRCh38
rs907584580 Chr.11: 85981790-85981790 on Build GRCh38
rs750029324 Chr.11: 85981773-85981773 on Build GRCh38
rs768605758 Chr.11: 85981840-85981840 on Build GRCh38
rs765804423 Chr.11: 85981802-85981802 on Build GRCh38
rs770940523 Chr.11: 85981733-85981733 on Build GRCh38

View Full Product Details