Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14853016-14853241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs747247621 Chr.X: 14853157-14853157 on Build GRCh38
rs776802337 Chr.X: 14853048-14853048 on Build GRCh38
rs1018343095 Chr.X: 14853077-14853077 on Build GRCh38
rs376384388 Chr.X: 14853076-14853076 on Build GRCh38
rs1057515805 Chr.X: 14853100-14853100 on Build GRCh38
rs765480337 Chr.X: 14853131-14853131 on Build GRCh38
rs758546007 Chr.X: 14853150-14853150 on Build GRCh38
rs763915979 Chr.X: 14853209-14853210 on Build GRCh38
rs776805534 Chr.X: 14853098-14853098 on Build GRCh38
rs762430531 Chr.X: 14853057-14853057 on Build GRCh38
rs759388298 Chr.X: 14853102-14853102 on Build GRCh38
rs755184176 Chr.X: 14853188-14853188 on Build GRCh38
rs752391089 Chr.X: 14853115-14853115 on Build GRCh38
rs748414147 Chr.X: 14853208-14853208 on Build GRCh38
rs781644458 Chr.X: 14853206-14853206 on Build GRCh38
rs762387195 Chr.X: 14853054-14853054 on Build GRCh38
rs759581894 Chr.X: 14853109-14853109 on Build GRCh38
rs772802668 Chr.X: 14853055-14853055 on Build GRCh38
rs753030842 Chr.X: 14853139-14853139 on Build GRCh38
rs750569208 Chr.X: 14853140-14853140 on Build GRCh38

View Full Product Details