Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72229927-72230188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7309767 c.921C>T p.I307I Chr.11: 72230001-72230001 on Build GRCh38
4146291 c.987A>G p.S329S Chr.11: 72230168-72230168 on Build GRCh38
5882127 c.856C>G p.L286V Chr.11: 72229936-72229936 on Build GRCh38
6225944 c.863A>T p.D288V Chr.11: 72229943-72229943 on Build GRCh38
7252541 c.909G>C p.K303N Chr.11: 72229989-72229989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17847215 Chr.11: 72229989-72229989 on Build GRCh38
rs771214975 Chr.11: 72230058-72230058 on Build GRCh38
rs2276048 Chr.11: 72230168-72230168 on Build GRCh38
rs368966021 Chr.11: 72230046-72230046 on Build GRCh38
rs1031911489 Chr.11: 72230044-72230044 on Build GRCh38
rs373511105 Chr.11: 72230005-72230005 on Build GRCh38
rs765778907 Chr.11: 72229984-72229984 on Build GRCh38
rs776507450 Chr.11: 72230065-72230065 on Build GRCh38
rs778042017 Chr.11: 72230029-72230029 on Build GRCh38
rs988183240 Chr.11: 72230084-72230084 on Build GRCh38
rs1038360832 Chr.11: 72230027-72230027 on Build GRCh38
rs375651667 Chr.11: 72229971-72229971 on Build GRCh38
rs749984847 Chr.11: 72230004-72230004 on Build GRCh38
rs764599724 Chr.11: 72230080-72230080 on Build GRCh38
rs749845241 Chr.11: 72230117-72230117 on Build GRCh38
rs770184208 Chr.11: 72230070-72230070 on Build GRCh38
rs767207284 Chr.11: 72230095-72230095 on Build GRCh38
rs371907420 Chr.11: 72229956-72229956 on Build GRCh38
rs754929406 Chr.11: 72230139-72230139 on Build GRCh38
rs202155977 Chr.11: 72229970-72229970 on Build GRCh38
rs375961815 Chr.11: 72230074-72230074 on Build GRCh38
rs367754263 Chr.11: 72229950-72229950 on Build GRCh38
rs111332650 Chr.11: 72230056-72230056 on Build GRCh38
rs751996555 Chr.11: 72230086-72230086 on Build GRCh38
rs763831147 Chr.11: 72229990-72229990 on Build GRCh38
rs375624221 Chr.11: 72230025-72230025 on Build GRCh38
rs765034075 Chr.11: 72229993-72229994 on Build GRCh38
rs780054338 Chr.11: 72230023-72230023 on Build GRCh38
rs755708388 Chr.11: 72230120-72230120 on Build GRCh38
rs370621872 Chr.11: 72230127-72230127 on Build GRCh38
rs759673818 Chr.11: 72230066-72230066 on Build GRCh38
rs763375013 Chr.11: 72230078-72230078 on Build GRCh38
rs761335284 Chr.11: 72230088-72230088 on Build GRCh38
rs780522806 Chr.11: 72230002-72230002 on Build GRCh38
rs879505794 Chr.11: 72229959-72229959 on Build GRCh38
rs372689513 Chr.11: 72230054-72230054 on Build GRCh38
rs779852630 Chr.11: 72230121-72230121 on Build GRCh38
rs370029165 Chr.11: 72229995-72229995 on Build GRCh38
rs997468345 Chr.11: 72230041-72230041 on Build GRCh38
rs200057735 Chr.11: 72229985-72229985 on Build GRCh38
rs753815166 Chr.11: 72230137-72230137 on Build GRCh38
rs751295151 Chr.11: 72229994-72229994 on Build GRCh38
rs201624140 Chr.11: 72229949-72229949 on Build GRCh38

View Full Product Details