Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34341889-34342162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOP10
Entrez Gene ID
55505
Gene Name
NOP10 ribonucleoprotein
Gene Aliases
DKCB1, NOLA3, NOP10P
UniGene
Hs.14317
Gene Chromosome Location
Chr.15: 34341716-34343161 on Build GRCh38
Transcripts
NM_018648.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4054145 c.82T>C p.C28R Chr.15: 34342081-34342081 on Build GRCh38
4468896 c.156C>T p.F52F Chr.15: 34342007-34342007 on Build GRCh38
4835955 c.66G>C p.P22P Chr.15: 34342097-34342097 on Build GRCh38
6727776 c.121T>C p.Y41H Chr.15: 34342042-34342042 on Build GRCh38
3377535 c.93T>C p.H31H Chr.15: 34342070-34342070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748193565 Chr.15: 34342074-34342074 on Build GRCh38
rs1045238 Chr.15: 34341923-34341923 on Build GRCh38
rs768707296 Chr.15: 34342069-34342069 on Build GRCh38
rs763635922 Chr.15: 34342012-34342012 on Build GRCh38
rs372176417 Chr.15: 34341924-34341924 on Build GRCh38
rs1042150335 Chr.15: 34341978-34341978 on Build GRCh38
rs121908092 Chr.15: 34342063-34342063 on Build GRCh38
rs780315311 Chr.15: 34342051-34342051 on Build GRCh38
rs777217218 Chr.15: 34341925-34341925 on Build GRCh38
rs201041278 Chr.15: 34341949-34341949 on Build GRCh38
rs773341436 Chr.15: 34342093-34342093 on Build GRCh38
rs377680031 Chr.15: 34341943-34341943 on Build GRCh38
rs936301020 Chr.15: 34342094-34342094 on Build GRCh38
rs768901406 Chr.15: 34341957-34341957 on Build GRCh38
rs779969910 Chr.15: 34342048-34342048 on Build GRCh38
rs375648407 Chr.15: 34341935-34341935 on Build GRCh38
rs199661222 Chr.15: 34342011-34342011 on Build GRCh38
rs1045204 Chr.15: 34341937-34341937 on Build GRCh38
rs422388 Chr.15: 34342123-34342123 on Build GRCh38
rs879535705 Chr.15: 34341937-34341938 on Build GRCh38
rs373843596 Chr.15: 34342071-34342071 on Build GRCh38
rs111425507 Chr.15: 34342111-34342111 on Build GRCh38
rs746979721 Chr.15: 34342061-34342061 on Build GRCh38
rs72720799 Chr.15: 34342117-34342117 on Build GRCh38
rs141981162 Chr.15: 34342119-34342119 on Build GRCh38
rs371001053 Chr.15: 34341966-34341966 on Build GRCh38
rs370853886 Chr.15: 34341936-34341936 on Build GRCh38
rs963339972 Chr.15: 34341915-34341915 on Build GRCh38
rs201604296 Chr.15: 34342029-34342029 on Build GRCh38
rs763236137 Chr.15: 34342000-34342000 on Build GRCh38
rs770460479 Chr.15: 34341934-34341934 on Build GRCh38
rs1045194 Chr.15: 34341938-34341938 on Build GRCh38
rs765367178 Chr.15: 34341984-34341984 on Build GRCh38
rs753180697 Chr.15: 34342142-34342142 on Build GRCh38
rs912544193 Chr.15: 34342090-34342090 on Build GRCh38
rs767899087 Chr.15: 34342138-34342138 on Build GRCh38
rs528879821 Chr.15: 34341926-34341927 on Build GRCh38
rs750603404 Chr.15: 34341988-34341988 on Build GRCh38
rs386782889 Chr.15: 34341937-34341938 on Build GRCh38
rs1013208714 Chr.15: 34341913-34341913 on Build GRCh38
rs776538895 Chr.15: 34341973-34341973 on Build GRCh38
rs562412129 Chr.15: 34342013-34342013 on Build GRCh38
rs368082745 Chr.15: 34342088-34342088 on Build GRCh38
rs762158306 Chr.15: 34341977-34341977 on Build GRCh38
rs758289839 Chr.15: 34342041-34342041 on Build GRCh38
rs200017156 Chr.15: 34342118-34342118 on Build GRCh38
rs756732260 Chr.15: 34342023-34342023 on Build GRCh38
rs766693193 Chr.15: 34342005-34342005 on Build GRCh38
rs745494123 Chr.15: 34342035-34342035 on Build GRCh38
rs763453630 Chr.15: 34342131-34342133 on Build GRCh38
rs193227 Chr.15: 34342109-34342109 on Build GRCh38
rs752083783 Chr.15: 34342009-34342009 on Build GRCh38
rs749018407 Chr.15: 34341928-34341928 on Build GRCh38

View Full Product Details