Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42752112-42752379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTBK2
Entrez Gene ID
146057
Gene Name
tau tubulin kinase 2
Gene Aliases
SCA11, TTBK
UniGene
Hs.646511
Gene Chromosome Location
Chr.15: 42738974-42920995 on Build GRCh38
Transcripts
NM_173500.3 XM_005254171.4 XM_005254173.4 XM_006720402.3 XM_006720403.3 XM_017021950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6810859 c.2979C>T p.G993G Chr.15: 42752267-42752267 on Build GRCh38
4737064 c.3082G>A p.V1028I Chr.15: 42752164-42752164 on Build GRCh38
1129203 c.3007C>G p.L1003V Chr.15: 42752239-42752239 on Build GRCh38
4570689 c.3068T>G p.F1023C Chr.15: 42752178-42752178 on Build GRCh38
396918 c.3035C>A p.A1012D Chr.15: 42752211-42752211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370550891 Chr.15: 42752282-42752282 on Build GRCh38
rs55735086 Chr.15: 42752186-42752186 on Build GRCh38
rs373433433 Chr.15: 42752273-42752273 on Build GRCh38
rs756527828 Chr.15: 42752215-42752215 on Build GRCh38
rs779236069 Chr.15: 42752236-42752236 on Build GRCh38
rs752316838 Chr.15: 42752211-42752211 on Build GRCh38
rs879080176 Chr.15: 42752329-42752329 on Build GRCh38
rs766921712 Chr.15: 42752327-42752327 on Build GRCh38
rs757774197 Chr.15: 42752232-42752232 on Build GRCh38
rs760324636 Chr.15: 42752346-42752346 on Build GRCh38
rs770922979 Chr.15: 42752302-42752302 on Build GRCh38
rs969467277 Chr.15: 42752287-42752287 on Build GRCh38
rs903031654 Chr.15: 42752209-42752209 on Build GRCh38
rs765923162 Chr.15: 42752349-42752349 on Build GRCh38
rs759127398 Chr.15: 42752320-42752320 on Build GRCh38
rs377620628 Chr.15: 42752266-42752266 on Build GRCh38
rs758922700 Chr.15: 42752198-42752198 on Build GRCh38
rs866877821 Chr.15: 42752350-42752350 on Build GRCh38
rs370728842 Chr.15: 42752267-42752267 on Build GRCh38
rs778104862 Chr.15: 42752221-42752221 on Build GRCh38
rs201662235 Chr.15: 42752225-42752225 on Build GRCh38
rs911630565 Chr.15: 42752165-42752165 on Build GRCh38
rs180791005 Chr.15: 42752305-42752305 on Build GRCh38
rs762984697 Chr.15: 42752155-42752155 on Build GRCh38
rs773343074 Chr.15: 42752153-42752153 on Build GRCh38
rs972275430 Chr.15: 42752355-42752355 on Build GRCh38
rs766166040 Chr.15: 42752169-42752169 on Build GRCh38
rs758376275 Chr.15: 42752258-42752258 on Build GRCh38
rs767231995 Chr.15: 42752203-42752203 on Build GRCh38
rs55919340 Chr.15: 42752207-42752207 on Build GRCh38
rs745988269 Chr.15: 42752253-42752253 on Build GRCh38
rs775188866 Chr.15: 42752337-42752337 on Build GRCh38

View Full Product Details