Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14859201-14859430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs200161949 Chr.X: 14859297-14859297 on Build GRCh38
rs1030317250 Chr.X: 14859399-14859399 on Build GRCh38
rs757806819 Chr.X: 14859237-14859237 on Build GRCh38
rs774030993 Chr.X: 14859380-14859380 on Build GRCh38
rs41309679 Chr.X: 14859282-14859282 on Build GRCh38
rs772102044 Chr.X: 14859356-14859358 on Build GRCh38
rs772510115 Chr.X: 14859371-14859372 on Build GRCh38
rs776590717 Chr.X: 14859381-14859381 on Build GRCh38
rs760500278 Chr.X: 14859387-14859390 on Build GRCh38
rs1057515807 Chr.X: 14859347-14859347 on Build GRCh38
rs941959108 Chr.X: 14859285-14859285 on Build GRCh38
rs771467972 Chr.X: 14859325-14859325 on Build GRCh38
rs773335115 Chr.X: 14859372-14859372 on Build GRCh38
rs746645943 Chr.X: 14859355-14859355 on Build GRCh38
rs986180195 Chr.X: 14859353-14859353 on Build GRCh38
rs139045247 Chr.X: 14859326-14859326 on Build GRCh38

View Full Product Details