Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27364713-27364982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B4
Entrez Gene ID
8890
Gene Name
eukaryotic translation initiation factor 2B subunit delta
Gene Aliases
EIF-2B, EIF2B, EIF2Bdelta
UniGene
Hs.169474
Gene Chromosome Location
Chr.2: 27364352-27370457 on Build GRCh38
Transcripts
NM_001034116.1 NM_001318965.1 NM_001318966.1 NM_001318967.1 NM_001318968.1 NM_001318969.1 NM_015636.3 NM_172195.3 XM_006712132.1 XM_011533147.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7078607 c.1375G>A p.E459K Chr.2: 27364775-27364775 on Build GRCh38
5468237 c.1351G>A p.A451T Chr.2: 27364739-27364739 on Build GRCh38
3734572 c.1238_1239insGGTGA p.S416fs*1 Chr.2: 27364851-27364852 on Build GRCh38
7078608 c.1315G>A p.E439K Chr.2: 27364775-27364775 on Build GRCh38
1251067 c.1337G>A p.R446H Chr.2: 27364753-27364753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748621364 Chr.2: 27364795-27364795 on Build GRCh38
rs780149885 Chr.2: 27364889-27364889 on Build GRCh38
rs1039565596 Chr.2: 27364935-27364935 on Build GRCh38
rs746154598 Chr.2: 27364751-27364751 on Build GRCh38
rs568549947 Chr.2: 27364954-27364954 on Build GRCh38
rs758540260 Chr.2: 27364752-27364752 on Build GRCh38
rs773426946 Chr.2: 27364800-27364800 on Build GRCh38
rs753573267 Chr.2: 27364872-27364872 on Build GRCh38
rs369616643 Chr.2: 27364813-27364813 on Build GRCh38
rs78115342 Chr.2: 27364945-27364945 on Build GRCh38
rs534043363 Chr.2: 27364914-27364914 on Build GRCh38
rs902546386 Chr.2: 27364960-27364960 on Build GRCh38
rs868522898 Chr.2: 27364787-27364787 on Build GRCh38
rs963221145 Chr.2: 27364933-27364933 on Build GRCh38
rs777400118 Chr.2: 27364740-27364740 on Build GRCh38
rs1058065 Chr.2: 27364857-27364857 on Build GRCh38
rs776776464 Chr.2: 27364908-27364910 on Build GRCh38
rs755135093 Chr.2: 27364894-27364894 on Build GRCh38
rs377146994 Chr.2: 27364807-27364807 on Build GRCh38
rs139134121 Chr.2: 27364891-27364891 on Build GRCh38
rs532964281 Chr.2: 27364933-27364935 on Build GRCh38
rs770188347 Chr.2: 27364917-27364917 on Build GRCh38
rs764012305 Chr.2: 27364861-27364861 on Build GRCh38
rs774024750 Chr.2: 27364939-27364939 on Build GRCh38
rs373193504 Chr.2: 27364918-27364918 on Build GRCh38
rs886055900 Chr.2: 27364780-27364780 on Build GRCh38
rs769203099 Chr.2: 27364934-27364936 on Build GRCh38
rs771280237 Chr.2: 27364921-27364921 on Build GRCh38
rs747532557 Chr.2: 27364815-27364815 on Build GRCh38
rs918271249 Chr.2: 27364865-27364865 on Build GRCh38
rs770318466 Chr.2: 27364797-27364797 on Build GRCh38
rs1042632344 Chr.2: 27364782-27364782 on Build GRCh38
rs759071387 Chr.2: 27364841-27364841 on Build GRCh38
rs767391188 Chr.2: 27364753-27364753 on Build GRCh38
rs535449285 Chr.2: 27364955-27364955 on Build GRCh38
rs777297913 Chr.2: 27364779-27364779 on Build GRCh38
rs759669141 Chr.2: 27364754-27364754 on Build GRCh38
rs747094092 Chr.2: 27364755-27364755 on Build GRCh38
rs757012537 Chr.2: 27364886-27364886 on Build GRCh38
rs372970925 Chr.2: 27364769-27364769 on Build GRCh38
rs781385220 Chr.2: 27364909-27364909 on Build GRCh38
rs756024921 Chr.2: 27364860-27364860 on Build GRCh38

View Full Product Details