Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181241443-181241690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
UniGene
Hs.5662
Gene Chromosome Location
Chr.5: 181236928-181243906 on Build GRCh38
Transcripts
NM_006098.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5128870 c.348C>T p.D116D Chr.5: 181241573-181241573 on Build GRCh38
5161554 c.283G>A p.G95S Chr.5: 181241638-181241638 on Build GRCh38
5204931 c.357G>T p.Q119H Chr.5: 181241564-181241564 on Build GRCh38
5749939 c.341C>A p.S114Y Chr.5: 181241580-181241580 on Build GRCh38
6346883 c.325C>A p.L109M Chr.5: 181241596-181241596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773028960 Chr.5: 181241497-181241497 on Build GRCh38
rs374020184 Chr.5: 181241516-181241516 on Build GRCh38
rs878893567 Chr.5: 181241489-181241490 on Build GRCh38
rs773788138 Chr.5: 181241541-181241541 on Build GRCh38
rs189371625 Chr.5: 181241481-181241481 on Build GRCh38
rs777527568 Chr.5: 181241653-181241653 on Build GRCh38
rs11540163 Chr.5: 181241555-181241555 on Build GRCh38
rs762269467 Chr.5: 181241601-181241601 on Build GRCh38
rs878915108 Chr.5: 181241485-181241485 on Build GRCh38
rs112143249 Chr.5: 181241640-181241640 on Build GRCh38
rs764463539 Chr.5: 181241549-181241549 on Build GRCh38
rs781177385 Chr.5: 181241651-181241651 on Build GRCh38
rs112513276 Chr.5: 181241490-181241490 on Build GRCh38
rs901614974 Chr.5: 181241486-181241487 on Build GRCh38
rs769607577 Chr.5: 181241495-181241495 on Build GRCh38
rs61751567 Chr.5: 181241646-181241646 on Build GRCh38
rs781220903 Chr.5: 181241467-181241467 on Build GRCh38
rs11540180 Chr.5: 181241588-181241588 on Build GRCh38
rs750877608 Chr.5: 181241627-181241627 on Build GRCh38
rs748931084 Chr.5: 181241510-181241510 on Build GRCh38
rs758636714 Chr.5: 181241639-181241639 on Build GRCh38
rs370922076 Chr.5: 181241642-181241642 on Build GRCh38
rs1050099239 Chr.5: 181241600-181241600 on Build GRCh38
rs759081911 Chr.5: 181241543-181241543 on Build GRCh38
rs748086030 Chr.5: 181241652-181241652 on Build GRCh38
rs368015408 Chr.5: 181241577-181241577 on Build GRCh38
rs111709706 Chr.5: 181241489-181241489 on Build GRCh38
rs75046714 Chr.5: 181241655-181241655 on Build GRCh38
rs755106833 Chr.5: 181241650-181241650 on Build GRCh38
rs781441209 Chr.5: 181241630-181241630 on Build GRCh38
rs11540186 Chr.5: 181241598-181241598 on Build GRCh38
rs765631889 Chr.5: 181241623-181241623 on Build GRCh38

View Full Product Details