Product Details

Assay Reference Genome
Location

Chr.X:132077640 on build GRCh38
Cytoband
Xq26.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
90167
Gene Symbol
FRMD7
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
Location
Chr.X:132076986-132128022 on build GRCh38
Assay Gene Location
Within Exon 13
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FRMD7 NM_001306193.1 12 2508 NP_001293122.1
NM_194277.2 12 2555 NP_919253.1
XM_017029947.1 12 2734 XP_016885436.1
XM_017029948.1 9 2196 XP_016885437.1
XM_017029949.1 8 2517 XP_016885438.1
AK125336.1 12 2555 BAC86135.1
AL161984.1 8 2217

View Full Product Details