Product Details
- Assay Reference Genome
Location
Chr.11:46645991 on build GRCh38
- Cytoband
- 11p11.2
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 9776
- Gene Symbol
- ATG13
- Gene Name
- autophagy related 13
- Gene Aliases
- KIAA0652, PARATARG8
- Location
- Chr.11:46617276-46676019 on build GRCh38
- Assay Gene Location
- Overlaps Exon 10 - Intron 10
View Full Product Details