Product Details
- Assay Reference Genome
Location
Chr.16:23854847 on build GRCh38
- Cytoband
- 16p12.2
- Species
- Homo sapiens
- Variation Type
- Copy Number
Additional Information
For this assay, SNP(s) [rs79214759] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Target Gene Details
- Entrez Gene ID
- 5579
- Gene Symbol
- PRKCB
- Gene Name
- protein kinase C beta
- Gene Aliases
- PKC-beta, PKCB, PRKCB1, PRKCB2
- Location
- Chr.16:23835979-24220611 on build GRCh38
- Assay Gene Location
- Within Intron 2
View Full Product Details