Product Details

Assay Reference Genome
Location

Chr.X:22055493 on build GRCh38
Cytoband
Xp22.11
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
5251
Gene Symbol
PHEX
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
Location
Chr.X:22032327-22251310 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
PHEX NM_000444.5 NP_000435.3
NM_001282754.1 NP_001269683.1
XM_017029579.1 XP_016885068.1
AD000712.1 AAB51604.1
AK298030.1
AK301528.1
BC105057.1 AAI05058.1
BC105059.1 AAI05060.1
U60475.1 AAC50552.1
U75645.1 AAB47749.1
U82970.1 AAC24487.1
U87284.1 AAB47562.1

View Full Product Details