Product Details

Assay Reference Genome
Location

Chr.8:27477370 on build GRCh38
Cytoband
8p21.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
1135
Gene Symbol
CHRNA2
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
Location
Chr.8:27459761-27479296 on build GRCh38
Assay Gene Location
Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
CHRNA2 NM_000742.3 NP_000733.2
NM_001282455.1 NP_001269384.1
XM_005273397.2 XP_005273454.1
XM_006716282.1 XP_006716345.1
XM_011544388.1 XP_011542690.1
XM_011544389.1 XP_011542691.1
XM_017013003.1 XP_016868492.1
XM_017013004.1 XP_016868493.1
AK296348.1
AK314787.1
DA407151.1
U62431.1 AAB40109.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv831275 Chr8:27470773 - 27624088 on Build GRCh38 Loss CHRNA2 EPHX2 MIR6843 CLU
nsv1020582 Chr8:27452251 - 27549327 on Build GRCh38 Gain CHRNA2 EPHX2 PTK2B

View Full Product Details