Product Details

Assay Reference Genome
Location

Chr.11:113419293 on build GRCh38
Cytoband
11q23.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

For this assay, SNP(s) [rs116658985] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Target Gene Details

Entrez Gene ID
1813
Gene Symbol
DRD2
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
Location
Chr.11:113409595-113475279 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
DRD2 NM_000795.3 NP_000786.1
NM_016574.3 NP_057658.2
XM_017017296.1 XP_016872785.1
AB209832.1 BAD93069.1
AB451480.1
AF176812.1 AAF61479.1
AK308777.1
BC021195.2 AAH21195.1
BF437994.1
M29066.1 AAA52761.1
M30625.1 AAA88024.1
S62137.1 AAB26819.1
S69899.1 AAB20571.1
X51362.1 CAA35746.1
X51645.1 CAB56463.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv556444 Chr11:113318201 - 113512672 on Build GRCh38 Gain TTC12 DRD2 ANKK1 MIR4301

View Full Product Details