Gene Transcripts

Gene Symbol
Mybpc1
Entrez Gene ID
109272
Gene Name
myosin binding protein C, slow-type
Gene Aliases
8030451F13Rik
Chromosome Location
Chr. 10: 88518279 - 88605234 on Build GRCm38
UniGene ID
Mm.147208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252372.1 NP_001239301.1 20, 21 2637
NM_175418.5 NP_780627.2 21, 22 2679
GenBank mRNA AK078761.1 - 1, 2 73
AK132188.1 - 20, 21 2510
BC062158.1 - 21, 22 2541 30134842

View Full Product Details