Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009654.3 | NP_033784.2 | 4 | 381 | |
GenBank mRNA | AJ011413.1 | - | 4 | 368 | |
AJ457860.1 | - | 4 | 335 | ||
AK010025.1 | - | 4 | 370 | ||
AK050248.1 | - | 4 | 370 | ||
AK050644.1 | - | 3 | 239 | ||
AK131891.1 | - | 4 | 373 | ||
AK160487.1 | - | 4 | 373 | ||
BC024643.1 | - | 4 | 355 | 4242076 | |
BC049971.1 | - | 4 | 355 | 4161986 | |
JX564908.1 | - | 4 | 335 | ||
JX564909.1 | - | 4 | 335 | ||
JX564910.1 | - | 4 | 335 | ||
M16111.1 | - | 1 | 41 |