Gene Transcripts

Gene Symbol
Smc2
Entrez Gene ID
14211
Gene Name
structural maintenance of chromosomes 2
Gene Aliases
5730502P04Rik, AI255214, AW545314, CAP-E, CAPE, Fin16, SMC-2, Smc2l1
Chromosome Location
Chr. 4: 52439221 - 52488365 on Build GRCm38
UniGene ID
Mm.2999
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301412.1 NP_001288341.1 20, 21 3025
NM_008017.4 NP_032043.3 20, 21 2949
GenBank mRNA AJ534939.1 - 19, 20 2780
AK145341.1 - 20, 21 2979
BC071232.1 - 5, 6 684
BC094380.1 - 20, 21 2921 30543190
U42385.1 - 3, 4 392

View Full Product Details