Gene Transcripts

Gene Symbol
Mrvi1
Entrez Gene ID
17540
Gene Name
MRV integration site 1
Gene Aliases
BB115629, Irag, Ris1
Chromosome Location
Chr. 7: 110868257 - 110982908 on Build GRCm38
UniGene ID
Mm.250023
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010826.5 NP_034956.2 16 2224
NM_194464.3 NP_919446.2 17 2467
GenBank mRNA AK141821.1 - 12 1506
AK157169.1 - 17 2077
U63407.1 - 17 2470
U63408.1 - 16 2219

View Full Product Details