Gene Transcripts

Gene Symbol
Scaf4
Entrez Gene ID
224432
Gene Name
SR-related CTD-associated factor 4
Gene Aliases
AA517739, Sfrs15, Sra4, Srsf15
Chromosome Location
Chr. 16: 90228951 - 90284498 on Build GRCm38
UniGene ID
Mm.439811
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178923.3 NP_849254.2 9, 10 1447
GenBank mRNA AK045266.1 - 9, 10 1444
AK173120.1 - 9, 10 1525
BC023734.1 - 6, 7 848
BC041782.1 - 6, 7 819
BC053096.1 - 9, 10 1342 6401143
BC057592.1 - 9, 10 1447 6417362

View Full Product Details