Gene Transcripts

Gene Symbol
Mpg
Entrez Gene ID
268395
Gene Name
N-methylpurine-DNA glycosylase
Gene Aliases
9830006D05, AI326268, APNG, Aag, Mid1
Chromosome Location
Chr. 11: 32226425 - 32232702 on Build GRCm38
UniGene ID
Mm.263161
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010822.3 NP_034952.2 3 593
GenBank mRNA AK018694.1 - 3 573
AK076201.1 - 3 588
AK088878.1 - 3 547
AK134900.1 - 4 930
AK155766.1 - 4 933
AK156067.1 - 3 560
BC014754.1 - 3 587 4486938
U10420.1 - 3 586

View Full Product Details