Gene Transcripts
- Gene Symbol
- Shank3
- Entrez Gene ID
- 58234
- Gene Name
- SH3/ankyrin domain gene 3
- Gene Aliases
- AI841104, SHANK3c-3, SHANK3c-4, Shank3b
- Chromosome Location
- Chr. 15: 89499707 - 89560261 on Build GRCm38
- UniGene ID
- Mm.146855
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021423.3 |
NP_067398.2
|
13, 14 |
1692 |
|
GenBank mRNA |
AB231013.1 |
- |
13, 14 |
1692 |
|
AB841411.1 |
- |
3, 4 |
656 |
|
AB841412.1 |
- |
3, 4 |
656 |
|
AJ245904.1 |
- |
11, 12 |
1546 |
|
HQ405759.1 |
- |
1, 2 |
123 |
|
HQ405760.1 |
- |
1, 2 |
123 |
|
HQ405764.1 |
- |
5, 6 |
394 |
|
HQ405765.1 |
- |
3, 4 |
257 |
|
HQ405766.1 |
- |
2, 3 |
143 |
|
KF612592.1 |
- |
6, 7 |
629 |
|
View Full Product Details