Gene Transcripts

Gene Symbol
Spesp1
Entrez Gene ID
66712
Gene Name
sperm equatorial segment protein 1
Gene Aliases
4921508E09Rik, AI428149
Chromosome Location
Chr. 9: 62270729 - 62282179 on Build GRCm38
UniGene ID
Mm.263615
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025721.2 NP_079997.1 2 324
GenBank mRNA AK014843.1 - 2 325
AK015620.1 - 2 355
BC050754.1 - 2 332 6773961

View Full Product Details