Gene Transcripts

Gene Symbol
Scarf1
Entrez Gene ID
380713
Gene Name
scavenger receptor class F, member 1
Gene Aliases
AA986099, SREC, SREC-I, mKIAA0149
Chromosome Location
Chr. 11: 75513495 - 75526580 on Build GRCm38
UniGene ID
Mm.10486
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004157.2 NP_001004157.2 1 63
GenBank mRNA AK133477.1 - 1 46
AK220215.1 - 1 47
BC057600.1 - 1 29
BC080288.1 - 1 63 6411836

View Full Product Details