Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271472.1 | NP_001258401.1 | 8 | 1052 | |
NM_001271473.1 | NP_001258402.1 | 5 | 719 | ||
NM_022723.3 | NP_073560.2 | 7 | 962 | ||
GenBank mRNA | AF276425.1 | - | 7 | 829 | |
AK041590.1 | - | 7 | 945 | ||
BC066066.1 | - | 8 | 1010 | 6850999 | |
BC080278.1 | - | 5 | 687 | 30354974 |