Gene Transcripts

Gene Symbol
Med8
Entrez Gene ID
80509
Gene Name
mediator complex subunit 8
Gene Aliases
2210021A15Rik, AB041805, ARC32
Chromosome Location
Chr. 4: 118409298 - 118415824 on Build GRCm38
UniGene ID
Mm.259567
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290688.1 NP_001277617.1 4 383
NM_020000.3 NP_064384.2 4 423
NM_173719.3 NP_776067.1 5 471
GenBank mRNA AB041805.1 - 3 1909
AK005422.1 - 5 454
AK008759.1 - 4 402
AK011080.1 - 4 383
AK147032.1 - 4 409
AK148025.1 - 4 404
BC002315.1 - 3 363
BC051683.1 - 4 361
BC053738.1 - 2 1270

View Full Product Details