Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163430.1 | NP_001156902.1 | 3 | 668 | |
NM_181586.3 | NP_853617.1 | 3 | 374 | ||
GenBank mRNA | AK013316.1 | - | 3 | 323 | |
AK145929.1 | - | 3 | 668 | ||
BC052763.1 | - | 3 | 324 | 6513375 |