Gene Transcripts

Gene Symbol
Slc13a5
Entrez Gene ID
237831
Gene Name
solute carrier family 13 (sodium-dependent citrate transporter), member 5
Gene Aliases
Indy, NaC2/NaCT, Nact, mINDY
Chromosome Location
Chr. 11: 72241990 - 72267032 on Build GRCm38
UniGene ID
Mm.340778
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004148.4 NP_001004148.1 10 1362
GenBank mRNA AK048662.1 - 10 1363
AK140943.1 - 10 1364
AK158253.1 - 10 1367
AY368893.1 - 10 1323

View Full Product Details