Gene Transcripts

Gene Symbol
March10
Entrez Gene ID
632687
Gene Name
membrane-associated ring finger (C3HC4) 10
Gene Aliases
4933417C16Rik, AV039223, OTTMUSG00000002847, Rnf190
Chromosome Location
Chr. 11: 105360798 - 105456863 on Build GRCm38
UniGene ID
Mm.121570
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039242.2 NP_001034331.1 3 407
NM_172568.2 NP_766156.2 3 407
GenBank mRNA AK132734.1 - 3 396
BC100474.1 - 3 400 30918723

View Full Product Details