Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025567.2 | NP_079843.1 | 4 | 549 | |
GenBank mRNA | AK002815.1 | - | 4 | 527 | |
AK011288.1 | - | 4 | 550 | ||
AK151299.1 | - | 4 | 540 | ||
AK170272.1 | - | 3 | 1149 | ||
AK186904.1 | - | 2 | 112 | ||
AK192516.1 | - | 4 | 406 | ||
BC005620.1 | - | 4 | 530 | 3711323 | |
BC082790.1 | - | 4 | 487 |