Gene Transcripts

Gene Symbol
Lrrc16a
Entrez Gene ID
68732
Gene Name
leucine rich repeat containing 16A
Gene Aliases
1110037D04Rik, AI425970, CARMIL, D130057M20, Lrrc16
Chromosome Location
Chr. 13: 24012481 - 24280790 on Build GRCm38
UniGene ID
Mm.211047
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026825.3 NP_081101.3 3, 4 386
GenBank mRNA AK051570.1 - 3, 4 386
AY437876.1 - 3, 4 391
BC150819.1 - 3, 4 312

View Full Product Details