Gene Transcripts

Gene Symbol
Ndufa11
Entrez Gene ID
69875
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 11
Gene Aliases
2010012C24Rik, AV006275, B14.7
Chromosome Location
Chr. 17: 56717762 - 56724248 on Build GRCm38
UniGene ID
Mm.279823
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027244.1 NP_081520.1 2 201
GenBank mRNA BC027690.1 - 2 150
BC053075.1 - 2 182
BC059729.1 - 2 180 6601702

View Full Product Details