Gene Transcripts

Gene Symbol
Sult6b1
Entrez Gene ID
73671
Gene Name
sulfotransferase family, cytosolic, 6B, member 1
Gene Aliases
2410078J06Rik, AV101767, ST6B1
Chromosome Location
Chr. 17: 78883938 - 78906992 on Build GRCm38
UniGene ID
Mm.45514
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163625.1 NP_001157097.1 3 374

View Full Product Details