Gene Transcripts

Gene Symbol
Sf3a3
Entrez Gene ID
75062
Gene Name
splicing factor 3a, subunit 3
Gene Aliases
4930512K19Rik, 60kDa
Chromosome Location
Chr. 4: 124714861 - 124732423 on Build GRCm38
UniGene ID
Mm.25779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029157.3 NP_083433.1 5 386
GenBank mRNA AK015776.1 - 5 365
AK030743.1 - 5 421
AK150733.1 - 5 384
AK151141.1 - 5 384
AK151382.1 - 5 384
AK161646.1 - 5 407
AK168966.1 - 5 410
AK169324.1 - 5 430
BC009141.1 - 5 386 3598647
BC092058.1 - 5 418 4017388

View Full Product Details